Cleanroom Labels
Polypropylene labels are clean, double-bagged, and heat-sealed
These polypropylene labels are ESD-safe and virtually free of particulates, leechables, and outgasses. With a service temperature of –20 to 176°F (–29 to 80°C) labels accept ink from cleanroom pens, thermal transfer, or direct thermal printers.
Removable labels pull easily and leave no residue. Each roll of 1000 labels is wound on a 3" ID polyethylene core.
Compliance: ISO Class 4 (formerly Class 10)
$179.75 - $230.59CAD / Roll of 1000
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (in) Width (in)
Availability Pricing
Texwipe ALB019BAAMT Cleanroom Labels, polypropylene, removable, 2" x 2" RK-86375-10 Mfr # ALB019BAAMT
COMPARE
2 2
$179.75CAD / Roll of 1000
Texwipe ALB164AAAMT Cleanroom Labels, polypropylene, removable, 3" x 2" RK-86375-15 Mfr # ALB164AAAMT
COMPARE
3 2
$230.59CAD / Roll of 1000