Spill-X 73834 Formaldehyde/Solvent Spill Treatment Kit
Representative image only.
Spill-X 73834 Formaldehyde/Solvent Spill Treatment Kit
Spill-X | Mfr: 73834| Item: 8629206
Treat chemical spills in the lab or on the plant floor with a single step!
$1,745.97CAD / Each
$1,745.97CAD / Each
Buy It Together
Current Item
Spill-X 73834 Formaldehyde/Solvent Spill Treatment Kit
Spill-X 73834 Formaldehyde/Solvent Spill Treatment Kit
8629206
$1,745.97CAD / Each
+
Spill-X 77265 Solvent Agent Refill, 6 containers
8629254
$1,687.03CAD / Pkg of 6
+
Spill-X 77255 Acid Agent Refill, 6 containers
8629250
$399.29CAD / Pkg of 6
+ =
Total Price 4744.32 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Spill Kit Treatment Guide, six polypropylene containers of agent, plastic case, one pair of rubber gloves and safety goggles, 100 pH test papers with color guide, scraper, two pick-up pans, and six disposal bags.
Key Features
  • Neutralizes hazardous liquid and chemical spills
  • Quick cleanups
  • 1 year warranty
More About this Item
These acid-neutralizing spill kits contain sorbents and other supplies which allow you to cleanup spills quickly. The easy-to-use shaker containers help neutralize hazardous liquid and chemical spills.
Order agent refills separately.
Spill-X-FP® Formaldehyde Polymerizer is formulated to rapidly treat formaldehyde spills and reduce their vapors to limit contamination. Reacts with aqueous formaldehyde to form a polynoxylin polymer.
Note: Where formaldehyde solution strength is less than 15%, apply Spill-X-S solvent adsorbent to solidify remaining liquid.
1 Year Warranty
Specifications
  • Chemical TreatedFormaldehyde/Solvent
  • Agent ColorGreen/Black
  • DescriptionFormaldehyde/Solvent Spill Treatment Kit