Sherwood Scientific 50035013 Tub assembly, non-sealed, 4 x 250 mL, glass

Representative image only.

Sherwood Scientific 50035013 Tub assembly, non-sealed, 4 x 250 mL, glass

Sherwood Scientific | Mfr: 50035013| Item: 0265862

Quickly and evenly dry particles as small as 5 μm

$1,194.92CAD / Each

$1,194.92CAD / Each

Buy It Together

Current Item

Sherwood Scientific 50035013 Tub assembly, non-sealed, 4 x 250 mL, glass
Sherwood Scientific 50035013 Tub assembly, non-sealed, 4 x 250 mL, glass
0265862

$1,194.92CAD / Each
+ + + + + =
Total Price 36081.12 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Faster and gentler than conventional drying cabinets—ideal for food products, polymers, soils, chemicals, pharmaceuticals, minerals, and waste materials
  • Variety of accessories provide easy customization for your drying applications

More About this Item

Non-Sealed Tub Assemblies are available in glass or stainless steel and require a filter bag, sold separately below.

Model 02658-02 includes a port to add extra sample and a port for a temperature/humidity probe (sold separately).

Order fluid bed dryer, filter bags, and optional accessories separately for a complete fluid bed dryer system.

Glass tubs are useful for observation of applications and determination of the optimum fluidizing velocity. Use multi-tub units for simultaneous drying of four 250-mL samples. Required filter bags and base sold separately.

Specifications
  • Materials of ConstructionGlass
  • DescriptionTub assembly, non-sealed, 4 x 250 mL, glass
Reviews
Q&A