SAS 18198-00 Microbial Air Sampler, Digital, Stainless Steel Head; 110-220 VAC
Representative image only.
SAS 18198-00 Microbial Air Sampler, Digital, Stainless Steel Head; 110-220 VAC
SAS | Mfr: 18198-20| Item: 3368702
Accurately collect and quantify airborne bacteria, viruses, molds, and fungi
$17,069.22CAD / Each
$17,069.22CAD / Each
Buy It Together
Current Item
SAS 18198-00 Microbial Air Sampler, Digital, Stainless Steel Head; 110-220 VAC
SAS 18198-00 Microbial Air Sampler, Digital, Stainless Steel Head; 110-220 VAC
3368702
$17,069.22CAD / Each
+ + =
Total Price 35800.43 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Rechargeable NiMH battery pack, battery charger, integrated RS-232 port for downloading data and carrying case.
NIST Calibration.
Key Features
  • Choose from eight preset sample volumes (10, 20, 30, 50, 100, 200, 500, or 1000 liters)
  • Program up to eight custom sample volumes
  • Detachable metal cover with intake holes creates laminar air flow
  • Flow rate is 100 liters/minute
  • Set parameters and view readings on the easy-to-read digital display
  • Unit retains previous sample volume
  • Microbes are driven onto the surface of a contact plate for incubation and analysis (order contact plate 14006-50 separately below)
  • 180l/min microbial air samplers are available, contact our application specialists for details
  • 1 year warranty
More About this Item
Impaction-style SAS air samplers let you accurately collect and quantify airborne bacteria, viruses, molds, and fungi. Ideal for clean rooms and process areas in pharmaceutical, electronics, and food tech environments. Microbes are driven onto the surface of a contact plate for incubation and analysis. Able to sample 45,000 liters of air per battery charge. 180l/min microbial air samplers are available, contact our application specialists for details.
Featuring a stainless steel head this sampler is ideal for clean room, pharmaceutical, and electronics applications. It meets material requirements for clean rooms. Includes a sturdy aluminum carrying/transport case.
1 Year Warranty
Specifications
  • MaterialStainless steel head
  • Height (in)4.125
  • Width (InchShort)12
  • Depth (InchShort)4 1/4
  • Battery1 Rechargeable NiMH (included)
  • Power (VAC)115;110 / 220
  • DescriptionMicrobial air sampler, digital, stainless steel head, 110-220 VAC