MAS 100 Air Sampler
Results you can trust in monitoring microbial air quality
  • Programmable sampling delay up to 59 minutes eliminating remote control devices
  • Use standard 100mm Petri dishes
The MAS 100 was developed on the Andersen Air Sampler Principle with collection apertures that correspond to Andersen Level 5 for excellent recovery of viable particles as small as 1µm. All models include a built-in calibration reminder that can be set between 1 and 12 months, and calibration is made easy with the new digital anemometer 39182-82 (sold separately). Internal software is easily accessed and controlled using single touch “yes or no?responses and pre-programmed volumes assure reproducible results.
Air flow is 100 liters/minute. The unit and lid cover are both made of anodized aluminum and the lids are autoclavable. Power is supplied via NiMH rechargeable batteries. The display is alpha-numeric LCD, 16 digits with real time and date.
MAS 100EcoTM Air Sampler
Microbiological air sampler for the food & beverage industry
Lightwight units offer an easy, low cost method for air monitoring
Handle doubles as a stand to allow different angles for testing
This model is specifically designed for the food and beverage industry. There are 6 preset volumes: 10, 20, 50, 100, 200 & 500 liters. Users can also reset any of the presets to any volume between 0-1000 liters. Run time for rechargeable NiMH batteries is 3 hours and maximum time for complete recharge is 9 hours.
MAS 100 Air Sampler
Samples up to 45,000 liters of air or 7.5 hours before battery needs to be recharged
This model offers a built-in anemometer compensates for different sampling heads, changes in temperature/pressure, and variably filled petri dishes. Collection volumes of 50, 100, 250, 500, and 1000 liters are preset for selection and can be deactivated for reactivated as needed. A specific volume between 0 and 2000 liters can also be selected.
$17,319.70 - $28,781.20CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
EMD Millipore 1.09227.0002 100 Eco Air Sampler, 100 L/min 3915280 Mfr # 1.09227.0001
COMPARE
100 Eco Air Sampler, 100 L/min
$17,319.70CAD / Each
EMD Millipore 1.09191.0001 MAS 100 NT Air Sampler and Microbial Air Monitoring System 3915288 Mfr # 1.09191.0001
COMPARE
MAS 100 NT Air Sampler and Microbial Air Monitoring System
$28,781.20CAD / Each