Mettler Toledo EG11 EasyPlus pH aqueous sensor
Representative image only.
Mettler Toledo EG11 EasyPlus pH aqueous sensor
Mettler Toledo | Mfr: 30043103| Item: 9419050
Enhance the versatility of your Mettler Toledo titrators
Call For Price
Call For Price
Buy It Together
Current Item
Mettler Toledo EG11 EasyPlus pH aqueous sensor
Mettler Toledo EG11 EasyPlus pH aqueous sensor
9419050
Call For Price
+ + =
Total Price 27861.80 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Aqueous sensor with cable and BNC connector
  • Used with Easy pH and Easy Pro titrators
  • 1 year warranty
More About this Item
Choose from a wide range of accessories compatible with your Mettler Toledo titrators for optimal performance.
Replacement for sensor included in kit.
1 Year Warranty
Specifications
  • Min pH (pH)0
  • Max pH (pH)14
  • mV Resolution0.1
  • DescriptionEasyPlus pH aqueous sensor