Labconco 9903200 Solvent Base Storage Cabinet for 30" Fume Hood

Representative image only.

Labconco 9903200 Solvent Base Storage Cabinet for 30" Fume Hood

Labconco | Mfr: 9903200| Item: 0911502

Provide support for your fume hood while adding additional storage

$6,821.80CAD / Each

$6,821.80CAD / Each

Buy It Together

Current Item

Labconco 9903200 Solvent Base Storage Cabinet for 30" Fume Hood
Labconco 9903200 Solvent Base Storage Cabinet for 30" Fume Hood
0911502

$6,821.80CAD / Each
+
Labconco 3746701 Telescoping Base Stand, 3 ft wide
0911471

$1,878.57CAD / Each
=
Total Price 8700.37 ADD 2 ITEMS TO CART
Product Description

Includes

Four leveling feet, one 8" filler panel to increase cabinet depth from 22" to 30", epoxy-coated steel shelf, and two rear vent connections with flame arrestors and closeure plugs.

Key Features

  • Choose between four types of storage cabinets – Acid, Solvent, Standard, and Vacuum Pump
  • Constructed of powder-coated steel
  • Supports loads of up to 800 pounds
  • Designed to efficiently store flammable materials and includes a 1-1/2" double wall construction for fire protection
  • Suitable for 30" fume hoods
  • 1 year warranty

More About this Item

Standard base cabinets are ideal for general storage of small laboratory equipment, mild acids ,and other reagents.

Acid storage cabinets allow you to safely store and vent acids and other corrosive liquids. The interior is constructed with a corrosion-resistant polyethylene liner to enhance corrosion resistance.

Solvent storage cabinets are suitable for storing and venting solvents and other flammable liquids. These cabinets will protect the interior contents in case of an external fire. These cabinets are constructed with dual wall powder-coated steel and feature a 1.5” air gap for fire protection.

Vacuum pump storage cabinets are utilized for vacuum pump storage.

Work surfaces are sold separately and required. Spillstopper work surfaces keep accidental spills and liquids away from the seam between the hood wall and surrounding countertop.

1 Year Warranty
Specifications
  • Hood Width (in)30
  • Exterior Width (in)30
  • Exterior Height (in)36.75
  • Exterior Depth (in)22
  • Shipping Weight (lbs)215
  • DescriptionSolvent Base Storage Cabinet for 30" Fume Hood
  • Warranty1 Year
Reviews
Q&A