Hach PHC10105 IntelliCAL pH Rugged Probe, 5-m cable
Representative image only.
Hach PHC10105 IntelliCAL pH Rugged Probe, 5-m cable
Hach | Mfr: PHC10105| Item: 5900474
Designed for Hach HQd meters only
$1,551.48CAD / Each
$1,551.48CAD / Each
Buy It Together
Current Item
Hach PHC10105 IntelliCAL pH Rugged Probe, 5-m cable
Hach PHC10105 IntelliCAL pH Rugged Probe, 5-m cable
5900474
$1,551.48CAD / Each
+ + +
Hach HQ1110 Portable Dedicated pH/ORP/mV Meter
5900500
$1,351.10CAD / Each
+
Hach HQ1130  Portable Dedicated Dissolved Oxygen Meter
5900505
$1,442.45CAD / Each
+ =
Total Price 21065.06 ADD 6 ITEMS TO CART
Product Description
Key Features
  • IntelliCAL Probes:
  • Digital solid-state technology for 'plug-and-play' performance
  • Factory calibrated-ready to use out of box
  • Move between meters without recalibrating
  • 1 year warranty
More About this Item
IntelliCAL probes store their own calibration data and identity. Double junction reference with ATC.
Rugged probes are IP68-rated with stainless-steel body, and are waterproof to 30-m for 24 hours
1 Year Warranty
Specifications
  • ApplicationspH
  • Cable Length (m)5
  • DescriptionIntelliCAL pH Rugged Probe, 5-m cable