Hach LBOD10101 IntelliCAL BOD LDO Probe, 1-m cable
Representative image only.
Hach LBOD10101 IntelliCAL BOD LDO Probe, 1-m cable
Hach | Mfr: LBOD10101| Item: 5900461
Designed for Hach HQd meters only
$2,585.80CAD / Each
$2,585.80CAD / Each
Buy It Together
Current Item
Hach LBOD10101 IntelliCAL BOD LDO Probe, 1-m cable
Hach LBOD10101 IntelliCAL BOD LDO Probe, 1-m cable
5900461
$2,585.80CAD / Each
+ + +
Hach HQ1110 Portable Dedicated pH/ORP/mV Meter
5900500
$1,351.10CAD / Each
+
Hach HQ1130  Portable Dedicated Dissolved Oxygen Meter
5900505
$1,442.45CAD / Each
+ =
Total Price 22099.38 ADD 6 ITEMS TO CART
Product Description
Key Features
  • IntelliCAL Probes:
  • Digital solid-state technology for 'plug-and-play' performance
  • Factory calibrated-ready to use out of box
  • Move between meters without recalibrating
  • 1 year warranty
More About this Item
IntelliCAL probes store their own calibration data and identity. Laboratory probes are waterproof to 3-m for 24 hours. Epoxy body.
1 Year Warranty
Specifications
  • ApplicationsDissolved Oxygen (BOD)
  • Cable Length (m)1
  • DescriptionIntelliCAL BOD LDO Probe, 1-m cable