Hach ISENH318101 IntelliCAL Ammonia ISE, 1-m cable
Representative image only.
Hach ISENH318101 IntelliCAL Ammonia ISE, 1-m cable
Hach | Mfr: ISENH318101| Item: 5900467
Designed for Hach HQd meters only
$1,771.01CAD / Each
$1,771.01CAD / Each
Buy It Together
Current Item
Hach ISENH318101 IntelliCAL Ammonia ISE, 1-m cable
Hach ISENH318101 IntelliCAL Ammonia ISE, 1-m cable
5900467
$1,771.01CAD / Each
+
Hach HQ1110 Portable Dedicated pH/ORP/mV Meter
5900500
$1,351.10CAD / Each
+ + + +
Hach HQ1130  Portable Dedicated Dissolved Oxygen Meter
5900505
$1,442.45CAD / Each
=
Total Price 21284.59 ADD 6 ITEMS TO CART
Product Description
Key Features
  • IntelliCAL Probes:
  • Digital solid-state technology for 'plug-and-play' performance
  • Factory calibrated-ready to use out of box
  • Move between meters without recalibrating
  • 1 year warranty
More About this Item
IntelliCAL probes store their own calibration data and identity. Double junction reference. Laboratory probes are waterproof to 3-m for 24 hours. Epoxy housing.
1 Year Warranty
Specifications
  • ApplicationsAmmonia
  • Cable Length (m)1
  • DescriptionIntelliCAL Ammonia ISE, 1-m cable