Fluke Calibration 2384-T Smart Connector, Thermocouple Probe (Blue Cap), Spare
Representative image only.
Fluke Calibration 2384-T Smart Connector, Thermocouple Probe (Blue Cap), Spare
Fluke Calibration | Mfr: 2384-T| Item: 9320260
Use to connect 1523 or 1524 reference thermometer to thermocouple leads
$381.61CAD / Each
$381.61CAD / Each
Product Description
Key Features
  • Supports reference junction compensation (RJC) with its own internal precision thermistor
  • Recommended for noble metal thermocouples such as Type S and Type R
  • 1 year warranty
More About this Item
Keep your thermometers running at their best with these accessories.
1 Year Warranty
Specifications
  • DescriptionSmart Connector, Thermocouple Probe (Blue Cap), Spare