EMD Millipore 1.09191.0001 MAS 100 NT Air Sampler and Microbial Air Monitoring System
Representative image only.
EMD Millipore 1.09191.0001 MAS 100 NT Air Sampler and Microbial Air Monitoring System
EMD Millipore | Mfr: 1.09191.0001| Item: 3915288
Results you can trust in monitoring microbial air quality
$28,781.20CAD / Each
$28,781.20CAD / Each
Buy It Together
Current Item
EMD Millipore 1.09191.0001 MAS 100 NT Air Sampler and Microbial Air Monitoring System
EMD Millipore 1.09191.0001 MAS 100 NT Air Sampler and Microbial Air Monitoring System
3915288
$28,781.20CAD / Each
+ =
Total Price 45850.42 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Programmable sampling delay up to 59 minutes eliminating remote control devices
  • Use standard 100mm Petri dishes
  • 1 year warranty
More About this Item
The MAS 100 was developed on the Andersen Air Sampler Principle with collection apertures that correspond to Andersen Level 5 for excellent recovery of viable particles as small as 1µm. All models include a built-in calibration reminder that can be set between 1 and 12 months, and calibration is made easy with the new digital anemometer 39182-82 (sold separately). Internal software is easily accessed and controlled using single touch “yes or no?responses and pre-programmed volumes assure reproducible results.
Air flow is 100 liters/minute. The unit and lid cover are both made of anodized aluminum and the lids are autoclavable. Power is supplied via NiMH rechargeable batteries. The display is alpha-numeric LCD, 16 digits with real time and date.
MAS 100EcoTM Air Sampler
Microbiological air sampler for the food & beverage industry
Lightwight units offer an easy, low cost method for air monitoring
Handle doubles as a stand to allow different angles for testing
This model is specifically designed for the food and beverage industry. There are 6 preset volumes: 10, 20, 50, 100, 200 & 500 liters. Users can also reset any of the presets to any volume between 0-1000 liters. Run time for rechargeable NiMH batteries is 3 hours and maximum time for complete recharge is 9 hours.
MAS 100 Air Sampler
Samples up to 45,000 liters of air or 7.5 hours before battery needs to be recharged
This model offers a built-in anemometer compensates for different sampling heads, changes in temperature/pressure, and variably filled petri dishes. Collection volumes of 50, 100, 250, 500, and 1000 liters are preset for selection and can be deactivated for reactivated as needed. A specific volume between 0 and 2000 liters can also be selected.
1 Year Warranty
Specifications
  • DescriptionMAS 100 NT Air Sampler and Microbial Air Monitoring System