Davis Instruments Anemometer for Vantage Pro2 and Vantage Pro
Representative image only.
Davis Instruments Anemometer for Vantage Pro2 and Vantage Pro
Davis Instruments | Mfr: 6410| Item: 3454264
Optimize performance of your Davis Instruments weather station
$435.02CAD / Each
$435.02CAD / Each
Buy It Together
Current Item
Davis Instruments Anemometer for Vantage Pro2 and Vantage Pro
Davis Instruments Anemometer for Vantage Pro2 and Vantage Pro
3454264
$435.02CAD / Each
+
Davis Instruments Anemometer/sensor transmitter kit
3000362
$940.61CAD / Each
+
Davis Instruments 6322C Cabled Integrated Sensor Suite
8640356
$1,503.01CAD / Each
=
Total Price 2878.64 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Compatible with different Davis Instruments weather stations
More About this Item
Get the most from your Davis Instruments weather stations with these accessories and components. Ensure the accessory is compatible with the weather station you have. Weather stations sold separately.
Replacement anemometer with mounting base for use with both wireless and cabled Vantage Pro2 and Vantage Pro weather stations.
Rugged contruction stands up to hurricane-force winds, yet is sensitive to light breezes. Sealed stainless steel bearings provides long life under harsh conditions.
Includes both wind speed and wind direction sensors, mounting hardware, and 40 ft. (12 m) cable.
Available for sale in the United States and Canada only.
Specifications
  • Max Wind Speed (mph)150
  • Min Wind Speed (mph)2
  • DescriptionAnemometer for Vantage Pro2 and Vantage Pro