Davis Instruments 7627 Solar-powered Repeater
Representative image only.
Davis Instruments 7627 Solar-powered Repeater
Davis Instruments | Mfr: 7627| Item: 8640343
Optimize performance of your Davis Instruments weather station
$1,486.72CAD / Each
$1,486.72CAD / Each
Buy It Together
Current Item
Davis Instruments 7627 Solar-powered Repeater
Davis Instruments 7627 Solar-powered Repeater
8640343
$1,486.72CAD / Each
+
Davis Instruments Anemometer/sensor transmitter kit
3000362
$940.61CAD / Each
+
Davis Instruments 6322C Cabled Integrated Sensor Suite
8640356
$1,503.01CAD / Each
=
Total Price 3930.34 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Compatible with different Davis Instruments weather stations
  • Used to extend distances or improve recepition,add one or more wireless repeaters. Range is up to 1000 ft line of sight or 200 to 400ft not line of sight.
  • 1 year warranty
More About this Item
Get the most from your Davis Instruments weather stations with these accessories and components. Ensure the accessory is compatible with the weather station you have. Weather stations sold separately.
Wireless Repeater receives data from any compatible transmitter and re-transmits it to any compatible Vantage Pro2 or Vantage Vue console/+receiver. The repeater extends the transmission range and improves reception between the transmitter and the receiver.
Each repeater has a transmitting and receiving range of between 200' to 1000' (75 to 300 m) depending upon the environment. The repeater can listen for up to eight different transmitter signals and can pass those signals on to another repeater or to any number of receivers.
Solar-powered repeaters (# 7627) include solar power panel, regulator, and battery.
Available for sale in the United States and Canada only.
1 Year Warranty
Specifications
  • DescriptionSolar-powered Repeater