Cole-Parmer Vacuum Filtration Holder; 47mm dia, 300 mL, sterility test

Representative image only.

Cole-Parmer Vacuum Filtration Holder; 47mm dia, 300 mL, sterility test

Cole-Parmer | Item: 0292310

Optimize your vacuum filtration process

$1,247.74CAD / Each

$1,247.74CAD / Each

Buy It Together

Current Item

Cole-Parmer Vacuum Filtration Holder; 47mm dia, 300 mL, sterility test
Cole-Parmer Vacuum Filtration Holder; 47mm dia, 300 mL, sterility test
0292310

$1,247.74CAD / Each
+ + + + + +
Cole-Parmer Filtering Flask; 2000 mL
3450922

$158.17CAD / Each
+ =
Total Price 7615.58 ADD 8 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Model 02923-20 meets EPA specifications for Hazardous Waste Toxicity Test
  • Autoclavable
  • Supplied with an aluminum clamp and a silicone rubber stopper
  • 1 year warranty

More About this Item

No. 5 stoppers fit standard 125-mL flasks; no. 8 stoppers fit 1-L flasks. Order no. 8B stopper 02924-02 separately at right to fit filter holders 02920-00 and 02921-00 to manifolds above. Filter holder 02923-10 is ideal for sterility testing of antibiotics and pharmaceuticals by bacteria retentive method described in USP XXI. This model includes funnel, filter base, aluminum clamp, and stopper. Model 02923-20 with sintered-glass membrane support meets EPA specifications for use in the Hazardous Waste Toxicity Test. Holder 02923-00, with stainless steel (SS) membrane support, provides ultraclean filtrate—screen does not shed particles.

Choose model 02923-10 for sterility testing of antibiotics and pharmaceutical by bacteria retentive method described in USP XXI. This model includes silicone funnel cover, borosilicate stopcock, silicone tubing for connection to sterile buffer, and SS vent needle with 13-mm SS filter holder.

1 Year Warranty
Specifications
  • Diameter (mm)47
  • Diameter (cm)4.7
  • Funnel Size (mL)300
  • Membrane SupportSintered Glass
  • Stopper NumberNo. 8
  • Filtration Area (cm²)9.6
  • DescriptionVacuum Filtration Holder; 47mm dia, 300 mL, sterility test
  • Warranty1 year
Reviews
Q&A