Advantec Vacuum Filtration Manifolds
Significantly reduce process time compared to gravity methods
  • Two- and three-way valves feature PTFE stopcocks for smooth control
  • Valves provide independent control at each branch for vacuum and venting
  • Connect three or six filter holders to one vacuum source
Stainless steel manifolds have two- or three-way PTFE valves for each branch to provide individual control of vacuum and air venting. Branches accept no. 8 stoppers. Hose barb accepts 3⁄8" ID vacuum tubing. Autoclavable.
PVC manifolds have branches with two-way PTFE valves for independent control of vacuum. Hose barb accepts 1⁄4" ID tubing. Branches accept no. 8 stoppers. Sanitize with ethanol or formaldehyde; do not autoclave. Max temperature is 140°F (60°C).
$2,851.01 - $6,954.40CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Branches Material Valve Type
Availability Pricing
Advantec 313000 Vacuum Filtration Manifold, Sterility Test; SS, 3-Branch with 3-Way Valves 0292400 Mfr # 313000
COMPARE
3 304 Stainless Steel 3-way
$4,272.83CAD / Each
Advantec 313200 Vacuum Filtration Manifold, Sterility Test; SS, 6-Branch with 3-Way Valves 0292410 Mfr # 313200
COMPARE
6 304 Stainless Steel 3-way
$6,954.40CAD / Each
Advantec 353100 Vacuum Filtration Manifold; Stainless Steel, 3-Branch with 2-Way Valves 0292420 Mfr # 353100
COMPARE
3 304 Stainless Steel 2-way
$3,624.54CAD / Each
Advantec 353300 Vacuum Filtration Manifold; Stainless Steel, 6-Branch with 2-Way Valves 0292430 Mfr # 353300
COMPARE
6 304 Stainless Steel 2-way
$6,048.27CAD / Each
Advantec 313400 Vacuum Filtration Manifold; PVC, 3-Branch with 2-Way Valves 0292440 Mfr # 313400
COMPARE
3 PVC 2-way
$2,851.01CAD / Each
Advantec 313600 Vacuum Filtration Manifold; PVC, 6-Branch with 2-Way Valves 0292450 Mfr # 313600
COMPARE
6 PVC 2-way
$4,353.87CAD / Each
Advantec 353130 Vacuum Filtration Manifold; Stainless Steel, 3-Branch with 3-Way Valves 0292485 Mfr # 353130
COMPARE
3 304 Stainless Steel 3-way
$3,116.22CAD / Each
Advantec 353330 Vacuum Filtration Manifold; Stainless Steel, 6-Branch with 3-Way Valves 0292495 Mfr # 353330
COMPARE
6 304 Stainless Steel 3-way
$5,871.46CAD / Each
Showing 1- 8 of 8