Cole-Parmer Vacuum Filtration Holder; 25mm; filtration area; 2.1 sq cm; funnel size; 15 mL

Representative image only.

Cole-Parmer Vacuum Filtration Holder; 25mm; filtration area; 2.1 sq cm; funnel size; 15 mL

Cole-Parmer | Item: 0292100

Optimize your vacuum filtration process

$455.28CAD / Each

$455.28CAD / Each

Buy It Together

Current Item

Cole-Parmer Vacuum Filtration Holder; 25mm; filtration area; 2.1 sq cm; funnel size; 15 mL
Cole-Parmer Vacuum Filtration Holder; 25mm; filtration area; 2.1 sq cm; funnel size; 15 mL
0292100

$455.28CAD / Each
+ + + + + + +
Cole-Parmer Filtering Flask; 2000 mL
3450922

$158.17CAD / Each
+ =
Total Price 6865.26 ADD 9 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Model 02923-20 meets EPA specifications for Hazardous Waste Toxicity Test
  • Autoclavable
  • Supplied with an aluminum clamp and a silicone rubber stopper
  • 1 year warranty

More About this Item

No. 5 stoppers fit standard 125-mL flasks; no. 8 stoppers fit 1-L flasks. Order no. 8B stopper 02924-02 separately at right to fit filter holders 02920-00 and 02921-00 to manifolds above. Filter holder 02923-10 is ideal for sterility testing of antibiotics and pharmaceuticals by bacteria retentive method described in USP XXI. This model includes funnel, filter base, aluminum clamp, and stopper. Model 02923-20 with sintered-glass membrane support meets EPA specifications for use in the Hazardous Waste Toxicity Test. Holder 02923-00, with stainless steel (SS) membrane support, provides ultraclean filtrate—screen does not shed particles.

To fit holder to manifold, order no. 8B stopper 02924-02 separately.

1 Year Warranty
Specifications
  • Diameter (mm)25
  • Diameter (cm)2.5
  • Funnel Size (mL)15
  • Membrane SupportSintered Glass
  • Stopper NumberNo. 5
  • Filtration Area (cm²)2.1
  • DescriptionVacuum Filtration Holder; 25mm; filtration area; 2.1 sq cm; funnel size; 15 mL
  • Warranty1 year
Reviews
Q&A