Cole-Parmer Standard Hazardous Location Refrigerator, Solid Door, 1°C to 10°C, 20 cubic ft, 120V

Representative image only.

Cole-Parmer Standard Hazardous Location Refrigerator, Solid Door, 1°C to 10°C, 20 cubic ft, 120V

Cole-Parmer | Item: 4426225

Reliable cold storage for hazardous environments

$8,180.27CAD / Each
$10,225.33CAD / Each

$8,180.27CAD / Each
$10,225.33CAD / Each

Buy It Together

Current Item

Cole-Parmer Standard Hazardous Location Refrigerator, Solid Door, 1°C to 10°C, 20 cubic ft, 120V
Cole-Parmer Standard Hazardous Location Refrigerator, Solid Door, 1°C to 10°C, 20 cubic ft, 120V
4426225

$8,180.27CAD / Each
$10,225.33CAD / Each
+ + + =
Total Price 20910.37 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Microprocessor temperature controller ensures accurate and stable conditions for critical samples from 1 to 10°C
  • Built for safety - Meets NFPA and OSHA guidelines for Hazardous Locations, Class 1, Division II, Group C & D
  • Advanced monitoring and security with digital display, audible/visual alarms, and remote alarm contacts for enhanced product protection
  • Environmentally friendly refrigerants, magnetic door gaskets, and cycle defrost for long-term reliability
  • 2 year warranty

More About this Item

Designed for use in hazardous locations, these refrigerators offer unparalleled temperature stability and security. Featuring a microprocessor temperature controller, alarm system, and remote monitoring capabilities, they provide the precision necessary for critical applications. Built to comply with NFPA and OSHA standards, these units ensure safe operation in volatile environments. With a rugged, hard-wired design and environmentally friendly refrigerants, they deliver both reliability and efficiency for demanding settings.

Specifications
  • Refrigerator Capacity (cu ft)20
  • Min Temperature - Refrigerator (° F)32
  • Max Temperature - Refrigerator (° F)50
  • Min Temperature - Refrigerator (° C)1
  • Max Temperature - Refrigerator (° C)10
  • Temperature ControlMicroprocessor
  • Refrigerator Interior Width (in)24 3/4
  • Refrigerator Interior Height (in)20 3/4
  • Refrigerator Interior Depth (in)65 1/4
  • Refrigerator Interior Width (cm)62.865
  • Refrigerator Interior Height (cm)52.705
  • Refrigerator Interior Depth (cm)165.735
  • Exterior Width (in)30 3/4
  • Exterior Height (in)74 5/8
  • Exterior Depth (in)29
  • Exterior Width (cm)78.105
  • Exterior Height (cm)189.5475
  • Exterior Depth (cm)73.66
  • Number of Doors1
  • Number of Shelves5
  • AlarmAudible and visual high and low temperature alarms
  • Cabinet ConstructionHigh density urethane foam cabinet construction
  • DisplayStandard Digital Temperature Display
  • Power (VAC)120
  • Power (Hz)60
  • Power (amps)2.5
  • DescriptionHazardous Location Refrigerator, Solid Door, 1°C to 10°C, 20 cubic ft, 120V
  • Warranty2 years
Documents & Certificates
Reviews
Q&A