Cole-Parmer® Standard Series Hazardous Location Refrigerators
Reliable cold storage for hazardous environments
  • Microprocessor temperature controller ensures accurate and stable conditions for critical samples from 1 to 10°C
  • Built for safety - Meets NFPA and OSHA guidelines for Hazardous Locations, Class 1, Division II, Group C & D
  • Advanced monitoring and security with digital display, audible/visual alarms, and remote alarm contacts for enhanced product protection
  • Environmentally friendly refrigerants, magnetic door gaskets, and cycle defrost for long-term reliability
Designed for use in hazardous locations, these refrigerators offer unparalleled temperature stability and security. Featuring a microprocessor temperature controller, alarm system, and remote monitoring capabilities, they provide the precision necessary for critical applications. Built to comply with NFPA and OSHA standards, these units ensure safe operation in volatile environments. With a rugged, hard-wired design and environmentally friendly refrigerants, they deliver both reliability and efficiency for demanding settings.
ITEM(S) ON SALE
$7,561.44 - $8,180.27CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Doors Power (VAC) Refrigerator Capacity (cu ft)
Availability Pricing
COMPARE
1 120 14
$7,585.02CAD / Each
$9,481.26CAD / Each
COMPARE
1 120 17
$7,561.44CAD / Each
$9,451.80CAD / Each
COMPARE
1 120 20
$8,180.27CAD / Each
$10,225.33CAD / Each