Cole-Parmer 500 PH 500 pH/Temperature 1/4-DIN Controller

Representative image only.

Cole-Parmer 500 PH 500 pH/Temperature 1/4-DIN Controller

Cole-Parmer | Item: 0580216

Isolated and reversible 4 to 20 mA output allow for proportional output, SPDT relays for On/Off control

$1,178.12CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$1,178.12CAD / Each

Buy It Together

Current Item

Cole-Parmer 500 PH 500 pH/Temperature 1/4-DIN Controller
Cole-Parmer 500 PH 500 pH/Temperature 1/4-DIN Controller
0580216

$1,178.12CAD / Each
+ + +
Cole-Parmer Epoxy pH Electrode BNC
0599223

$259.54CAD / Each
+ + + + + =
Total Price 5451.46 ADD 9 ITEMS TO CART
Product Description

Key Features

  • Graphic LCD for clear measurement display
  • Automatic buffer recognition of US and NIST buffers
  • Automatic temperature compensation
  • Password protected ensures security of system
  • RS-485 output for remote access allows multiple unit control from your computer
  • Backlit diplay
  • 1 year warranty

More About this Item

Cole-Parmer online controllers offer features found in more expensive models. Graphic LCD simultaneously displays measurement, temperature, electrode status, relay status, and transmitter current output. Calibration data is stored in memory; making the unit ready for measurement. Password protection, RS-485 for remote access, and the isolated and reversible 4 to 20 mA output offer extended data options with security.

The pH 500 (05802-16) controller has one isolated and reversible 4 to 20 mA output and five programmable on/off relays: four for pH and one for temperature--ideal for high/low control to activate an alarm, a pump, or another device. ATC element is a 10 kilohm thermistor.

1 Year WarrantyConformite EuropeenneIP 65
Specifications
  • Control TypeOn/Off
  • Display TypeLCD
  • Housing Material1/4-DIN
  • Min pH (pH)0
  • Max pH (pH)16
  • pH Resolution0.01
  • pH Accuracy±1% + 1 digit
  • Min mV (mV)0
  • Min Temperature (° C)-10
  • Max Temperature (° C)120
  • Min Temperature (° F)14
  • Max Temperature (° F)248
  • Temperature Resolution0.1°
  • Temperature Accuracy±0.3°C
  • Temperature CompensationAutomatic
  • Password ProtectionYes
  • Output Current4-20mA (Reversible)
  • Output Relay5 SPDT relays
  • Power (VAC)115 / 230
  • Power (Hz)50/60
  • Width (cm)9.53
  • Height (cm)9.53
  • Width (in)3 3/4
  • Height (in)3 3/4
  • Depth (in)5 13/16
  • Description500 pH/Temperature 1/4-DIN Controller
  • Warranty1 year
Reviews
Q&A