Product Type
Bed Mass (mg)
Industry
Kinesis Microplate Products
Kinesis has a variety of microplates for you to select from including the popular TELOS® Microplate™ plate which is optimized for the sample processing and extraction of small-volume biological fluids. The internal diameter, frit design, and sorbent allow biological fluid samples to be processed. A modular design of the microplate allows for flexibility in sample numbers. Full- or partially-populated plates can be processed using vacuum or positive pressure. In addition, the well outlet design ensures good collection plate penetration, removing any possibility of well-to-well cross contamination.
Kinesis Telos Microplates — Optimized Well Design
  • • The SPE sorbent has been moved into the tip, optimizing particle size and frit porosity for reliable sample processing.
    • Narrow tip design provides optimum bed height. 50 µL elution volumes minimize analyte dilution/evaporation times.
    • Populate base plate with choice of TELOS neo SPE Sorbents to support analyte focused method development.
  • Kinesis Telos Microplate Optimized Well Design
  • • Cylindrical well — no sample creep or cross-contamination.
    • The modular design allows for flexibility in sample numbers preventing waste of partially used plates.
    • The well outlet design ensures good collection plate penetration, removing any possibility of cross-contamination between wells.
Chromatography Supplies and Consumables