Corning® Stripwell™ Plates
Flat-bottom strips break apart and can be reassembled into strip holder
  • Strips have one-way fit into strip holder—prevents misorientation
  • Manufactured from polystyrene (PS)— ideal for uniform binding, high optical clarity, and low background absorption
The Stripwell™ plates and strips are 1 x 8 well flat bottom strips designed to easily break apart to be placed into strip holder. Plates include strips and strip holder preassembled in a 96-well format. Strips are sold without a strip holder, holder 13020-50 sold separately. High and medium binding surface options available for a wide range of applications.
High binding surface binds medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions— ideal for immunoassays. Binding capacity is approximately 400 to 500ng IgG/cm².
Medium binding surface is hydrophobic in nature and binds biomolecules through passive interactions and is suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface. Binding capacity is approximately 100 to 200ng IgG/cm².
$947.39 - $1,580.95CAD / Case of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Binding Property Plate Type Description
Availability Pricing
Corning Stripwell™ Plates, 96-Well, Polystyrene, High Binding; 100/Cs 1302000 Mfr # 2592
COMPARE
High Plate Plates, 96-Well, Polystyrene, High Binding; 100/Cs
$1,580.95CAD / Case of 100
Corning Stripwell™ Strips, 1 x 8 Well Strips, Polystyrene, High Binding; 800/Cs 1302012 Mfr # 2580
COMPARE
High Strip Strips, 1 x 8 Well Strips, Polystyrene, High Binding; 800/Cs
$947.39CAD / Case of 800