Overcoming Viscosity Challenges with Peristaltic Pumps
Viscous fluids are often essential to critical processes but can be notoriously difficult to handle. Whether dealing with thick pharmaceutical suspensions, sticky food ingredients, or high-viscosity chemical blends, pumping these fluids without clogging, contamination, or flow disruption requires the correct equipment. Peristaltic pumps offer a robust, non-contact solution for moving viscous materials with precision and reliability.
Ice Cream coming out of the Ice Cream machine
Understanding Viscosity in Fluid Processes
Viscosity is a fluid’s resistance to flow. As viscosity increases, so does the difficulty in transferring the fluid through a pump system. Highly viscous fluids are prone to:
  • Reduced flow rates
  • Excessive back pressure
  • Cavitation in non-positive displacement pumps
  • Increased wear on pump components
Inaccurate handling of these fluids can lead to downtime, product loss, or contamination in sensitive environments. That’s why equipment selection and pump design matters.
Why Peristaltic Pumps Excel with Viscous Fluids
Peristaltic pumps move fluids by compressing and releasing a flexible tube using rotating rollers. This positive displacement action creates a vacuum that draws the fluid into the tubing and pushes it forward. Because the fluid only contacts the tubing and not the pump mechanism itself, peristaltic pumps offer several key advantages for viscous applications:
1. Self-Priming and Dry-Running
Peristaltic pumps are inherently self-priming and can handle suction lift without additional components. They can also run dry without damage, which can be helpful when fluids slowly feed due to high viscosity.
2. Seal-Free, Clog-Free Design
There are no valves, seals, or diaphragms to clog, an important benefit when dealing with pastes, gels, or slurries that can jam in such components.
3. Gentle, Low-Shear Handling
The squeezing action provides a smooth, low-shear flow, protecting sensitive fluids (e.g., cell suspensions, emulsions, protein-based formulations) from degradation.
4. High Accuracy and Repeatability
Each rotor rotation delivers a fixed volume of fluid, making flow consistent even as viscosity varies. When properly calibrated, peristaltic pumps maintain this accuracy across various fluid types.
5. Easy to Clean and Maintain
The fluid path is limited to the tubing, which is easy to replace or change sizes and is ideal for hygienic or multiproduct environments.
Peristaltic Solutions for High-Viscosity Applications
Cole-Parmer offers a wide range of peristaltic pumps engineered to handle challenging fluids. Whether you need precise dosing or bulk transfer, there’s a model built for your application.
Watson-Marlow 323 Series Pumps
The Watson-Marlow 323 peristaltic pumps are intuitive and feature a modular, interchangeable pump head design. The digital control allows for precise speed regulation, which is crucial when handling viscous fluids that require slower, controlled flow rates to prevent tubing stress.
Why they work for viscous fluids:
  • Modular design accepts multiple and interchangeable pump heads, providing flexibility to switch tubing sizes or adjust flow rates easily
  • Self-priming and reversible—ideal for tricky startups and complete drainage
Watson-Marlow 530 Series Pumps
The 530 Series peristaltic pumps offer greater torque and pumping capacity, making them ideal for handling more viscous materials. The built-in calibration mode ensures accurate flow rates even when fluid resistance is high, allowing you to dose thick or non-Newtonian fluids with greater precision and confidence.
Why they work for viscous fluids:
  • Higher torque capability handles increased resistance from thicker media
  • Compatible with larger bore and thicker wall tubing to accommodate higher flow rates and pressure
  • Built-in calibration and MemoDose operations provide enhanced accuracy and ease of pumping
Tubing
Your peristaltic pump is only as effective as the tubing inside it. Tubing must offer the right combination of strength, flexibility, and chemical compatibility when handling viscous fluids to ensure consistent flow and long-term performance. Cole-Parmer offers a wide range of tubing formulations designed to handle challenging applications.
  • Cole-Parmer® Black TPE Tubing: Designed for long-lasting performance, this thermoplastic elastomer (TPE) tubing is an excellent choice for pumping oils or surfactants in chemical and industrial applications. Engineered to resist abrasion and flexural fatigue, this tubing offers excellent dimensional stability, helping to reduce maintenance in continuous-use systems.
  • PharMed® BPT Biocompatible Peristaltic Pump Tubing: Engineered for biopharmaceutical applications, PharMed BPT tubing offers exceptional flex life, a key advantage when pumping viscous media over extended periods. It resists fatigue and maintains performance in systems requiring sterility or regulatory compliance.
  • Tygon® E-LFL Long Flexible Life Tubing: For applications where tubing wear is a concern, this tubing delivers one of the longest pumping lives available while maintaining precise, repeatable flow rates.
  • Tygon® 2475 IB High Pressure Thermoplastic Tubing: High-viscosity fluids can generate significant back pressure, especially in closed systems. With an inner braid construction, this tubing maintains integrity under pressure, which is ideal when flow resistance is high and tubing collapse is a concern.
  • Watson-Marlow Bioprene® TPE Pump Tubing: This TPE tubing is designed specifically for use in Watson-Marlow peristaltic pumps. It offers a low-sorption, low-spallation fluid path and resists degradation over long production cycles.
Pump on a lab bench
Key Considerations When Pumping Viscous Fluids
System setup must be carefully aligned with the fluid's viscosity and process needs to ensure optimal performance. Consider the following:
1. Tubing Selection
Viscous fluids require tubing with high mechanical strength to resist collapse under suction and maintain consistent occlusion. To suit these conditions, choose larger bore, thicker wall, or reinforced variants.
2. Drive Power and Pump Head Configuration
Thicker fluids require higher torque and lower speeds for optimal performance. Models like the Watson-Marlow 323 Series feature interchangeable pump heads to allow multiple channels, while the 530 Series is equipped to deliver the torque needed, larger tubing bore size, and thicker wall for better flow rates.
3. Flow Rate and Calibration
Flow rates are impacted by viscosity, especially at lower pump speeds. It's recommended that the system be calibrated using built-in pump modes, gravimetric methods, or volumetric displacement to ensure accurate dosing. Utilize the MemoDose feature on select Watson-Marlow pumps to accurately dispense a set quantity each time.
4. Temperature and Pressure
In some applications, pre-heating a fluid can reduce viscosity for easier transfer. However, care must be taken not to exceed the temperature or pressure limits of the tubing and pump. Operating within manufacturer guidelines ensures longevity and safety.
Application Examples
Peristaltic pumps are trusted across industries to handle viscous fluids without sacrificing process integrity:
  • Pharmaceutical and Biotech: Pumping gel-based formulations or viscous excipients under sterile, closed-loop conditions.
  • Food & Beverage: Accurate dosing of syrups, sauces, honey, or flavoring concentrates, with full washdown compatibility.
  • Chemical Manufacturing: Transferring adhesives, polymers, or resins that would clog traditional pumps.
  • Cosmetics: Filling and transferring lotions, creams, and serums without introducing air or contamination.
Cosmetic Cream coming out of the machine
Tips for Success
  • Always prime tubing before operation to avoid dry friction.
  • Operate at lower speeds when pumping high-viscosity fluids to minimize tubing wear.
  • Monitor tubing for signs of fatigue and replace on a preventive schedule.
  • Secure tubing properly within the pump head and verify correct occlusion settings during setup.
Conquer Viscosity with Confidence
Viscous fluids don’t have to slow down your process. With the right peristaltic pump and tubing combination, you can move thick, sticky, or shear-sensitive materials with precision and confidence. Cole-Parmer offers a comprehensive portfolio of peristaltic pumping solutions to meet the unique challenges of viscous fluid handling.
Explore our full range of peristaltic pumps and tubing, or connect with a fluid handling specialist for expert guidance on your high-viscosity application.