Tygon® E-LFL Long Flexible Life Tubing
Designed specifically for use in peristaltic pump applications
  • Extremely low particle spallation
  • Long flex life – reduces downtime
  • Suitable for high purity applications
  • Meets USP Class VI, NSF-51 and FDA criteria
  • Notice: Tygon® E LFL is not compliant with applicable European Union (EU) and Japan food and beverage regulations. Do not use this product for applications subject to these standards in these regions.
Compared to other clear tubing, the Tygon® E-LFL tubing offers longer pump life at 0 psi as well as at back pressure up to 25 psi during peristaltic pump operations. Because of its long flex life the tubing is able to reduce production downtime due to pump tubing failure in laboratory applications.
The reduction of particle spallation eliminates the need for frequent downstream filter replacements and lessens the risk of sensitive-fluid contamination. The tubing is manufactured with non-DEHP bio-based plasticizers which increase pump performance while reducing environmental impact. The tubing is also resistant to gas permeation compared to silicone, helping to protect your sensitive fluids from oxidation or adulteration.
Applications include a wide range of liquid transfer in labs, surfactant delivery, food and cosmetic processing, bioprocessing, food dispensing, and shear-sensitive fluid transfer. (Note: do not use for food and beverage applications in the EU or Japan.)
$247.53 - $797.10CAD / Pkg of 1
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Tygon S3 E-LFL Tubing, 1/16" ID x 3/16" OD; 25 Ft 5011355 Mfr # AVX42003
COMPARE
1/16 3/16 1/16
$247.53CAD / Pkg of 1
Tygon S3 E-LFL Tubing, 1/8" ID x 1/4" OD; 25 Ft 5011356 Mfr # AVX42007
COMPARE
1/8 1/4 1/16
$286.21CAD / Pkg of 1
Tygon S3 E-LFL Tubing, 3/16" ID x 5/16" OD; 25 Ft 5011357 Mfr # AVX42012
COMPARE
3/16 5/16 1/16
$301.68CAD / Pkg of 1
Tygon S3 E-LFL Tubing, 1/4" ID x 3/8" OD; 25 Ft 5011358 Mfr # AVX42017
COMPARE
1/4 3/8 1/16
$286.21CAD / Pkg of 1
Tygon S3 E-LFL Tubing, 1/4" ID x 1/2" OD; 25 Ft 5011359 Mfr # AVX42019
COMPARE
1/4 1/2 1/8
$503.16CAD / Pkg of 1
Tygon S3 E-LFL Tubing, 5/16" ID x 7/16" OD; 25 Ft 5011360 Mfr # AVX42022
COMPARE
5/16 7/16 1/16
$340.35CAD / Pkg of 1
Tygon S3 E-LFL Tubing, 3/8" ID x 5/8" OD; 25 Ft 5011361 Mfr # AVX42029
COMPARE
3/8 5/8 1/8
$758.80CAD / Pkg of 1
Tygon S3 E-LFL Tubing, 1/2" ID x 3/4" OD; 25 Ft 5011362 Mfr # AVX42038
COMPARE
1/2 3/4 1/8
$797.10CAD / Pkg of 1
Tygon S3 E-LFL Tubing, 3/4" ID x 1- 1/4" OD; 10 Ft 5011363 Mfr # AVX06057
COMPARE
3/4 1 1/4 1/4
$696.18CAD / Pkg of 1
Tygon S3 E-LFL Tubing, 1" ID x 1- 3/8" OD; 10 Ft 5011364 Mfr # AVX06064
COMPARE
1 1 3/8 3/16
$766.16CAD / Pkg of 1
Showing 1- 10 of 10