Jenway 74 & 76 Spectrophotometer Installation and Setup
Jenway 74 & 76 Spectrophotometer tutorials: Installation Setup | Performing Modes | Extra Features
Summary
Equipped with CPLive™ technology, the 74 & 76 Series comprises Jenway’s most intuitive and user-friendly spectrophotometers yet. The Jenway 74 & 76 Series Spectrophotometers are easy to unpack and install. Once turned on the units undertake self-checks, after which you can easily access the settings to configure the auto-save function and favourites, in addition to inputting your time, date and language.
What’s in the box?
The box contains 74 or 76 spectrophotometer UK, EU & US power leads and a power supply unit.
Getting set up
  1. Plug in the power supply unit
  2. Plug in an ethernet cable (not supplied), to gain access to the network
  3. Turn the unit on and wait for self-checks to be performed
  4. Once the main menu appears go into the settings menu and set:
    1. Time
    2. Language
    3. Date
    4. Measurement settings (including auto-save)
  5. Check
    1. Network connection and CPLive connection
    2. Storage space
Please note that service settings are for service engineers only
Find out more about CPLive
Browse all Jenway Spectrophotometers and Accessories