Jenway Spectrophotometers and Accessories
Designed with your convenience in mind, Jenway life science products offer many innovative features to help you become more efficient and productive. When you choose Jenway, you are ensured robust and reliable data analysis. Front and center of this product family are the innovative Jenway spectrophotometers. These easy-to-use devices provide simple set up and fast navigation with intuitive touchscreen displays. The newer touchscreens, like on the 74 Series spectrophotometers, are fast and responsive even with gloves. CPLive connectivity allow you to keep your data secure and easily accessible with CPLive connectivity—results are automatically uploaded and stored in the cloud. You can transfer and store all your spectrophotometry protocols and results to one place, and effortlessly share with colleagues. Simultaneously manage multiple devices through your free CPLive account, via a tablet or computer. Or, use the CPLive app on your smartphone. Choose from a wide selection of spectrometers, Visible/UV/Visible spectrophotometers, life science analyzers, fluorometers, colorimeters, conductivity meters, pH meters, flame photometers, oxygen metters, cuvettes, and accessories.
What is CPLive?
Share data and manage your account with 24/7 remote access from anywhere.
• CPLive connectivity keeps your data secure and easily accessible.
• Results are automatically uploaded and stored in the cloud.
• Transfer and store all your spectrophotometry protocols and results to one place, and effortlessly share with colleagues.
• Simultaneously manage multiple devices through your free CPLive account, via a tablet or computer.
• Use the CPLive app on your smartphone.
Jenway Spectrophotomers Offer Efficiency, Accuracy and Versatility
Compare models to identify the spectrophotometer that best meets your needs. Jenway Specifications Comparison Chart
Getting a Handle on Temperature
VISIBLE, UV/VIS Spectroscopy: Single & Split Beam Overview
Getting a Handle on Temperature
Spectroscopy Applications
Jenway 74 Series Visible UV-Vis Scanning Spectrophotometers with CPLive
72 Series
• Diode array technology
• Simultaneous measurement of all wavelengths, scan time as short as 3 seconds
• Color touch screen and user-friendly interface
Jenway 74 Series Visible UV-Vis Scanning Spectrophotometers with CPLive
74 Series
• Single Beam Technology
• The Jenway 7415 Nano measures the purity and concentration of 0.5µl-5µl biological sample volumes.
Synthesis, Workup, Purification products and supplies
76 Series
• Split Beam Technology
• 3.6GB internal memory
• CPLive cloud-based secure storage of your results and protocol
Need Help with this product?
Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.
Call Us At800-363-5900
Live chat