Zefon Preloaded Air Sampling Cassettes with Glass Fiber Membrane
Fully assembled and preloaded with filter membrane for quick and easy sampling
  • Leak-free design—eliminates sample loss
  • Preloaded with 37-mm diameter glass fiber membrane
  • Clear styrene housing provides excellent visibility of sample
  • Conform to most NIOSH standards
Make air sampling fast and simple with preassembled, preloaded filter cassettes. Glass fiber membrane contains no binders or strengthening agents, making it ideal for suspended solid sampling and atomic absorption spectroscopy. Glass fiber allows fine particle retention while exhibiting high flow rates and high load capacity.
$127.82 - $131.43CAD / Box of 50
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Diameter (mm) Pore Size (µm) Description
Availability Pricing
COMPARE
1.0 Cassette, Preloaded, 37 mm, 2 Piece, Styrene, 1.0 µm, Glass Fiber; 50/BX
$131.43CAD / Box of 50
COMPARE
1.0 Cassette, Preloaded, 37 mm, 3 Piece, Styrene, 1.0 µm, Glass Fiber; 50/BX
$127.82CAD / Box of 50