Yamato Programmable Mechanical Convection Ovens
Self-diagnostic function alerts you to sensor or heater malfunctions
  • Excellent temperature accuracy
  • PID temperature controller provides easy program settings
  • Oven calibration functions are ideal for validation work
  • Simultaneously view set point and actual temperatures
  • Built-in exhaust ports help remove moisture
  • Keypad lock prevents accidental setting changes
  • Easy-to-clean stainless steel interior
These programmable mechanical convection ovens provide excellent uniformity, autotune, calibration offset, and zero-span calibration. Easy operation features include quick auto-stop, auto-stop, auto-start, and programmed auto-start. Program up to 30 steps (a combination of set temperature and set time) for your operation. Self-diagnostic functions alert you both visually and audibly if sensor or heater malfunctions; heater circuit automatically turns off. Specially designed large push-button door handle ensures smooth, stress-free opening and closing of oven door. Add an external temperature probe using the 33 mm dia cable port.
$5,722.65 - $13,543.40CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Max Temperature (° C) Power (VAC)
Availability Pricing
Yamato DKN-402 Programmable Mechanical Convection Oven, 3.2 Cu Ft, 115VAC 3395905 Mfr # DKN-402C
COMPARE
90 260 115
$5,722.65CAD / Each
Yamato DKN-812 Programmable Mechanical Convection Oven, 10.7 Cu Ft, 220 VAC 3395907 Mfr # DKN-812C
COMPARE
303 260 220
$13,543.40CAD / Each