WTW pH 3310 ProfiLine Handheld Meter Only

Representative image only.

WTW pH 3310 ProfiLine Handheld Meter Only

WTW | Mfr: 2AA310| Item: 5889020

A rugged new line of electrochemistry meters with a range of features, benefits, and price points to meet every application and budget!

$1,482.23CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$1,482.23CAD / Each

Buy It Together

Current Item

WTW pH 3310 ProfiLine Handheld Meter Only
WTW pH 3310 ProfiLine Handheld Meter Only
5889020

$1,482.23CAD / Each
+ + =
Total Price 2078.58 ADD 3 ITEMS TO CART
Product Description

Key Features

  • Advanced 3310 models feature an innovative waterproof USB connection for ultra-flexible data output
  • Broad pH buffer recognition to meet most calibration needs
  • Rugged, waterproof housing is great for the lab or the field
  • 3 year warranty

More About this Item

Durable meters feature a 100% waterproof and easy-to-clean keypad. Silicone rubber keys provide tactile feedback for error-free operation. The large graphic display allows for excellent readability even under difficult lighting conditions. A built-in calibration timer ensures more reliable results.

pH 3310 meters add extended data memory for logging up to 5000 GLP-compliant data sets. Time-controlled datalogging function offers adjustable data collection intervals—ideal for long-term field monitoring. A special “sleep” mode saves power and extends battery life when performing long-term monitoring. Nonvolatile-memory saves all stored data even if battery power is lost. Data transfer to a PC is made simple via the unique waterproof mini-USB interface.

Meters include: four AA batteries.

Meter kits add: specified probe, calibration solutions, and hard carrrying case.

Meters with drinking water kit further add: Sentix® 81 probe, stand, beaker, measuring vessel, and CD-ROM.

3 Year WarrantyConformite EuropeenneIP 67ISO9001:2000 Certified Supplier
Specifications
  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.005 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.3 mV + 1 digit
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • Temperature compensationAutomatic or manual
  • InterfaceUSB
  • WaterproofYes
  • Battery4 x AA batteries (included)
  • Width (cm)5.595874
  • Height (cm)7.897622
  • Width (in)2 13/64
  • Length (in)7
  • Height (in)3 7/64
  • DescriptionProfiLine Handheld Meter Only
  • Warranty3 year
Reviews
Q&A