Wet/Wet Digital Differential Pressure Gauges
Rugged, intrinsically safe, ATEX certified gauge
  • High-accuracy, long-term vacuum measurement
  • 15 and 100 PSI versions rated to 100 PSI static pressures
  • Customizable for panel mounting or portable configurations
  • Programmable for custom engineering units
  • Optional datalogging capability
  • LCD backlit display
  • Easy to use buttons
The XP2i-DP is a very rugged, intrinsically safe, ATEX certified gauge that has a 0.1% of reading accuracy and is capable of use over continuous vacuum. Gauges with software provide an integrated data logging solution for your applications. The differential gauge is a combination of test gauge technology, double-sided isolated differential pressure sensing, and rugged industrial design. The Wet/Wet differential transducer features the ability to measure fluids. The differential pressure sensor is a double-sided, media compatible, piezeoresistive silicon pressure sensor packaged in a stainless steel housing. The sensing packages utilizes oil to transfer pressure from the two stainless steel diaphragms to the sensing element. It is designed for high performance, low pressure applications. The stainless steel package makes it suitable for use with liquids and corrosive gases.
Applications:
Gas filter testing
Level measurement
Rotary gas meter testing
Autoclave vacuum monitor / calibration
Available in the USA only.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Crystal AX15PSIXP2I-DP-DL Digital Differential Gauge, +/-15 PSI With Data Logging Software 6806404 Mfr # 15PSIXP2I-DP-DL
COMPARE
±0.02% full-scale <20% full scale, ±0.1% of reading >20% full scale 15