Weighing Funnels, Glass
Safe transfer of samples to volumetric flasks
  • No-roll, flat side for placement on balance
  • Reusable
These reusable borosilicate glass weighing funnels are designed to ensure complete sample transfer to volumetric flasks. Funnels are flat on one side so you can set them on a balance. Micro funnel is sized to fit microwell plates.
$177.18 - $270.00CAD / Pkg of 6
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Stem OD (mm)
Availability Pricing
COMPARE
Glass 7
$177.18CAD / Pkg of 6
COMPARE
Glass 8
$182.70CAD / Pkg of 6
COMPARE
Glass 11
$210.69CAD / Pkg of 6
COMPARE
Glass 17
$250.84CAD / Pkg of 6
COMPARE
Glass 20
$270.00CAD / Pkg of 6