Watson-Marlow Pumpsil® Platinum-Cured Silicone Pump Tubing
Ultra-smooth bore minimizes protein binding and bacterial growth
  • Exceptionally pure tubing—ideal for biopharmaceutical industry
  • Manufactured to achieve high precision, consistency, and durability
  • Fully traceable—laser etched part number, lot number, and use-by date on each tubing
  • Wide operating temperature range of –20 to 80°C (–4 to 176°F)
  • Manufactured in an ISO 114644-1 Class 7 Cleanroom
  • Sterilize by autoclave, EtO, or gamma irradiation
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Pumpsil platinum-cured silicone tubing provides the precision, consistency, and durability needed for worry-free pumping. Each tubing comes with the part number, lot number, and use-by-date laser-etched along the length of the tube, ensuring full traceability even when the tubing is removed from the bag.
The platinum-cured tubing is free of the 2,4 DCBA residue as well as phthalate and BPA-free, making this an ideal choice for the biopharmaceutical industry. The ultra-smooth bore helps minimize protein binding, ensuring high-purity finished product. Tubing is manufactured and double-packed in heat-sealed polyethylene bags in an ISO Class 7 cleanroom to ensure that the integrity is not compromised.
$126.86 - $444.96CAD / Pkg of 1
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (m) Tubing ID (mm) Wall Thickness (mm)
Availability Pricing
Watson-Marlow Pumpsil Platinum-Cured Silicone Pump Tubing, 0.5 mm Bore x 1.6 mm Wall; 5 m 5013022 Mfr # 913.AJ05.016
COMPARE
5 0.5 1.6
$127.89CAD / Pkg of 1
Watson-Marlow Pumpsil Platinum-Cured Silicone Pump Tubing, 0.8 mm Bore x 1.6 mm Wall; 5 m 5013023 Mfr # 913.AJ08.016
COMPARE
5 0.8 1.6
$126.86CAD / Pkg of 1
Watson-Marlow Pumpsil Platinum-Cured Silicone Pump Tubing, 1.6 mm Bore x 1.6 mm Wall; 5 m 5013024 Mfr # 913.AJ16.016
COMPARE
5 1.6 1.6
$145.28CAD / Pkg of 1
Watson-Marlow Pumpsil Platinum-Cured Silicone Pump Tubing, 3.2 mm Bore x 1.6 mm Wall; 5 m 5013025 Mfr # 913.AJ32.016
COMPARE
5 3.2 1.6
$174.60CAD / Pkg of 1
Watson-Marlow Pumpsil Platinum-Cured Silicone Pump Tubing, 4.8 mm Bore x 1.6 mm Wall; 5 m 5013026 Mfr # 913.AJ48.016
COMPARE
5 4.8 1.6
$205.17CAD / Pkg of 1
Watson-Marlow Pumpsil Platinum-Cured Silicone Pump Tubing, 6.4 mm Bore x 1.6 mm Wall; 5 m 5013357 Mfr # 913.AJ64.016
COMPARE
5 6.4 1.6
$209.22CAD / Pkg of 1
Watson-Marlow Pumpsil Platinum-Cured Silicone Pump Tubing, 8.0 mm Bore x 1.6 mm Wall; 5 m 5013358 Mfr # 913.AJ80.016
COMPARE
5 8 1.6
$235.74CAD / Pkg of 1
Watson-Marlow Pumpsil Platinum-Cured Silicone Pump Tubing, 2.4 mm Bore x 1.6 mm Wall; 5 m 5013359 Mfr # 913.AJ24.016
COMPARE
5 2.4 1.6
$153.97CAD / Pkg of 1
Watson-Marlow Pumpsil Platinum-Cured Silicone Pump Tubing, 1.6 mm Bore x 2.4 mm Wall; 5 m 5013366 Mfr # 913.AJ16.024
COMPARE
5 1.6 2.4
$251.95CAD / Pkg of 1
Watson-Marlow Pumpsil Platinum-Cured Silicone Pump Tubing, 3.2 mm Bore x 2.4 mm Wall; 5 m 5013367 Mfr # 913.AJ32.024
COMPARE
5 3.2 2.4
$257.84CAD / Pkg of 1
Watson-Marlow Pumpsil Platinum-Cured Silicone Pump Tubing, 4.8 mm Bore x 2.4 mm Wall; 5 m 5013368 Mfr # 913.AJ48.024
COMPARE
5 4.8 2.4
$260.79CAD / Pkg of 1
Watson-Marlow Pumpsil Platinum-Cured Silicone Pump Tubing, 6.4 mm Bore x 2.4 mm Wall; 5 m 5013369 Mfr # 913.AJ64.024
COMPARE
5 6.4 2.4
$297.62CAD / Pkg of 1
Watson-Marlow Pumpsil Platinum-Cured Silicone Pump Tubing, 8.0 mm Bore x 2.4 mm Wall; 5 m 5013376 Mfr # 913.AJ80.024
COMPARE
5 8 2.4
$368.35CAD / Pkg of 1
Watson-Marlow Pumpsil Platinum-Cured Silicone Pump Tubing, 9.6 mm Bore x 2.4 mm Wall; 5 m 5013377 Mfr # 913.AJ96.024
COMPARE
5 9.6 2.4
$444.96CAD / Pkg of 1
Showing 1- 14 of 14