Water Bath / Magnetic Stirrer Systems
Simple setup, easy-to-grip handles, and efficient design
  • Precise temperature and stirring control
  • Use as water bath, stirrer bath, or heat evaporator for various applications
Use baths and stirrers separately or together. Bath components incorporate durable 304 SS inner construction with 316L SS heating element. Baked-on melamine finish on galvanized steel bath exterior. Over-current protection fuse and empty-boiling preventative safety devices. Stirrers feature a powerful rare-earth magnet with smooth speed control to a maximum speed of 1500 rpm. Easy-to-clean design, the main body is made of durable glass fiber-reinforced polybutylene terephthalate, which is heat- and chemical-resistant. Combine baths and magnetic stirrers (sold separately) for excellent temperature uniformity and stirring action. Typical applications include protein digestion and polymer research.
$1,377.62 - $1,915.41CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Reservoir Capacity (L)
Availability Pricing
Advantec TBS181SB Bath for Bath/Magnetic Stirrer Systems, 3.8 L, 120 VAC/60 Hz 0291801 Mfr # TBS181SB
COMPARE
95 115 3.8
$1,377.62CAD / Each
Advantec TBS221SB Bath for Bath/Magnetic Stirrer Systems, 8 L, 120 VAC/60 Hz 0291805 Mfr # TBS221SB
COMPARE
95 115 8
$1,448.34CAD / Each
Advantec Bath for Bath/Magnetic Stirrer Systems, 8 L, 220 VAC/50 Hz 0291807 Mfr # TBS221SB(220V)
COMPARE
95 230 8
$1,448.34CAD / Each
Advantec TBS271SB Bath for Bath/Magnetic Stirrer Systems, 12 L, 120 VAC/60 Hz 0291809 Mfr # TBS271SB
COMPARE
95 115 12
$1,915.41CAD / Each