Vibro-Separators
Ideal for large batch quantities
  • Product contacts only polished stainless steel parts
  • Provides noiseless, high-frequency vibrations for efficient sifting and particle separations
These versatile separators are a must for anyone who needs to screen or scalp powders and grains, or sift particles from liquid while on site. Use these separators in laboratories, bakeries, mills, or on-site.
Separators feature an 18'' diameter upper frame, and a bottom frame with a 6'' open discharge spout. A four-pole drive motor provides silent three dimensional vibration. Quick-release clamps allow easy disassembly for cleaning and sifting different products. All parts that come in contact with product are made of polished stainless steel (SS).
$9,886.45 - $10,159.02CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Shipping Weight (kg) Description
Availability Pricing
CE International Trading Vibro-Separator Nonportable, SS Model, 115 VAC 5999810 Mfr # LS-450-S-115V/60HZ
COMPARE
115 63.5 Nonportable, SS Model, 115 VAC
$9,886.45CAD / Each
CE International Trading Vibro-Separator Portable, SS Model, 230 VAC 5999835 Mfr # LS-450-SA-230V/50HZ CE MARKED
COMPARE
230 72.6 Portable, SS Model, 230 VAC
$10,159.02CAD / Each