Versilon® C-210-A Oil, Fuel, and Grease Resistant Tubing
Tough ether-base polyurethane tubing offers exceptional resistance to oils, grease, and fuels
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
  • Excellent abrasion and tear resistance
  • Remains flexible even in subzero temperatures
  • Consistently tight lot-to-lot tolerances
  • Clear tubing—view fluid being transferred
The Versilon® C-210-A tubing provides exceptional stability when exposed to oils, fuels, and grease. Use this tough ether-based polyurethane tubing for transferring abrasive and viscous slurries, dispensing lubricants and degreasers, and recovering coolants. The plasticizer-free tubing offers excellent chemical resistance and withstands temperatures ranging from -73 to 79°C (-100 to 175°F).
$68.81 - $1,395.30CAD / Pkg of 1
25 variations of this product are available.
See All Product Options Below
25 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 1/16" ID x 1/8" OD; 50 Ft 5010689 Mfr # AEM02002
COMPARE
1/16 1/8 1/32
$68.81CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 1/8" ID x 3/16" OD; 50 Ft 5010690 Mfr # AEM02006
COMPARE
1/8 3/16 1/32
$85.16CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 1/8" ID x 1/4" OD; 50 Ft 5010691 Mfr # AEM02007
COMPARE
1/8 1/4 1/16
$162.44CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 3/16" ID x 1/4" OD; 50 Ft 5010692 Mfr # AEM02011
COMPARE
3/16 1/4 1/32
$119.20CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 3/16" ID x 5/16" OD; 50 Ft 5010693 Mfr # AEM02012
COMPARE
3/16 5/16 1/16
$208.85CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 3/16" ID x 3/8" OD; 50 Ft 5010694 Mfr # AEM02013
COMPARE
3/16 3/8 3/32
$510.53CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 1/4" ID x 5/16" OD; 50 Ft 5010695 Mfr # AEM02016
COMPARE
1/4 5/16 1/32
$216.59CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 1/4" ID x 3/8" OD; 50 Ft 5010696 Mfr # AEM02017
COMPARE
1/4 3/8 1/16
$255.26CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 1/4" ID x 7/16" OD; 50 Ft 5010697 Mfr # AEM02018
COMPARE
1/4 7/16 3/32
$495.06CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 1/4" ID x 1/2" OD; 50 Ft 5010698 Mfr # AEM02019
COMPARE
1/4 1/2 1/8
$665.24CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 5/16" ID x 7/16" OD; 50 Ft 5010699 Mfr # AEM02022
COMPARE
5/16 7/16 1/16
$363.56CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 3/8" ID x 1/2" OD; 50 Ft 5010700 Mfr # AEM02027
COMPARE
3/8 1/2 1/16
$301.68CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 3/8" ID x 9/16" OD; 50 Ft 5010701 Mfr # AEM02028
COMPARE
3/8 9/16 3/32
$464.12CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 3/8" ID x 5/8" OD; 50 Ft 5010702 Mfr # AEM02029
COMPARE
3/8 5/8 1/8
$680.71CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 7/16" ID x 5/8" OD; 50 Ft 5010703 Mfr # AEM02033
COMPARE
7/16 5/8 3/32
$603.35CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 7/16" ID x 11/16" OD; 50 Ft 5010704 Mfr # AEM02034
COMPARE
7/16 11/16 1/8
$758.80CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 1/2" ID x 5/8" OD; 50 Ft 5010705 Mfr # AEM02036
COMPARE
1/2 5/8 1/16
$448.65CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 1/2" ID x 11/16" OD; 50 Ft 5010706 Mfr # AEM02037
COMPARE
1/2 11/16 3/32
$642.40CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 1/2" ID x 3/4" OD; 50 Ft 5010707 Mfr # AEM02038
COMPARE
1/2 3/4 1/8
$820.68CAD / Pkg of 1
Versilon C-210-A Oil, Fuel, and Grease Resistant Tubing, 5/8" ID x 3/4" OD; 50 Ft 5010708 Mfr # AEM02044
COMPARE
5/8 3/4 1/16
$541.47CAD / Pkg of 1
Showing 1 - 20 of 25