Velp Flocculator Jar Testers

Save money and chemicals by testing on a small scale
  • ASTM D2035 compliant method
  • Single motor drives all positions at a consistent speed
  • Choose from portable or laboratory models, with four or six positions
Jar testers are designed to test the efficiency of flocculation or precipitation reagents on a small scale. Stirring rod shafts can individually be stopped and height can be adjusted while the system is operating. Choose from portable or laboratory testers.

$3,354.02 - $6,279.51CAD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Number of Positions Stirring Control Timer Control
Availability Pricing
Velp Fp4-Portable FP4 Portable 4-position Flocculator 100-240V 9956105 Mfr # F105A0117
COMPARE
4 20, 40, 50, 100, 120 RPM 1 to 30 min
$3,354.02CAD / Each
Velp F105A0108 JLT4 Laboratory 4-Position Flocculator 100-240V 9956113 Mfr # F105A0108
COMPARE
4 10 to 300 RPM 1 to 999 min, 1 to99 hrs, Continuous
$4,560.73CAD / Each
Velp F105A0109 JLT6 Laboratory 6-Position Flocculator 100-240V 9956115 Mfr # F105A0109
COMPARE
6 10 to 300 RPM 1 to 999 min, 1 to 99hrs, Continuous
$5,212.04CAD / Each
Velp F105A0111 FC4S 4-Position Flocculator 100-240V 9956117 Mfr # F105A0111
COMPARE
4 10, 15, 30, 45, 60, 90, 120, 150, 200 RPM Continuous
$5,128.50CAD / Each
Velp F105A0112 FC6S 6-Position Flocculator 100-240V 9956119 Mfr # F105A0112
COMPARE
6 10, 15, 30, 45, 60, 90, 120, 150, 200 RPM Continuous
$6,279.51CAD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat