Velp F105A0111 FC4S 4-Position Flocculator 100-240V

Representative image only.

Velp F105A0111 FC4S 4-Position Flocculator 100-240V

Velp | Mfr: F105A0111| Item: 9956117

Save money and chemicals by testing on a small scale

$5,128.50CAD / Each

$5,128.50CAD / Each

Buy It Together

Current Item

Velp F105A0111 FC4S 4-Position Flocculator 100-240V
Velp F105A0111 FC4S 4-Position Flocculator 100-240V
9956117

$5,128.50CAD / Each
+ + =
Total Price 13801.02 ADD 3 ITEMS TO CART
Product Description

Includes

stainless steel stirring rods and universal AC adapter.

Key Features

  • ASTM D2035 compliant method
  • Single motor drives all positions at a consistent speed
  • Choose from portable or laboratory models, with four or six positions
  • Each position has independent stirring controls
  • 2 year warranty

More About this Item

Jar testers are designed to test the efficiency of flocculation or precipitation reagents on a small scale. Stirring rod shafts can individually be stopped and height can be adjusted while the system is operating. Choose from portable or laboratory testers.

Laboratory Jar Tester is available with four positions with a digital display that shows stirring speed and time. the FC model offers individual position speed control at 10, 15, 30, 45, 60, 90,120, 150, 200 rpm. Featuring an illuminated back panel for enhanced visibility. Laboratory units require standard 2-L beakers (order separately,34512-13 4/pk).

Two year warrantyConformite Europeenne
Specifications
  • Meter TypeBenchtop
  • Power (VAC)100 to 240
  • Power (Hz)50 or 60
  • Width (cm)64.77
  • Height (cm)34.29
  • Width (in)25 1/2
  • Length (in)10 1/4
  • Height (in)13 1/2
  • Number of Positions4
  • Stirring Control10, 15, 30, 45, 60, 90, 120, 150, 200 RPM
  • Timer ControlContinuous
  • DescriptionFC4S 4-Position Flocculator 100-240V
Reviews
Q&A