Velp ARE 5 Analog Stirring Hot Plates
Offers consistent stirring and heating for general lab applications
  • Can reach maximum temperature of 310 °C (590 °F)
  • Stirring speed up to 1500 rpm
  • Able to stir up to 15 liters
  • Two illuminated indicators warn when heating is on
  • IP42 certified
  • 2 year warranty
The easy-to-use ARE 5 analog stirring hot plates are designed to be safe and offer you powerful stirring and excellent heating for basic industrial and academic laboratory applications. The units are safe to use. A hot plate warning icon illuminates when the temperature is above 50 °C (122 °F). Internal electronic parts are protected by the insulated disk and die-cast aluminum housing.
The 135 mm diameter aluminum alloy top heating plate ensures temperature homogeneity and optimum heat distribution across the entire surface. The plate is resistant to thermal stress and shock.
The powerful brushless motor can stir up to 15 liters from 100 to 1500 rpm. SpeedServo™ torque compensation technology ensures you constant speed as viscosity changes.
$900.24CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
High Speed (rpm) Max Stirring Volume (Liters) Power (VAC)
Availability Pricing
Velp ARE 5 Analog Stirring Hot Plate, Aluminum; 115V 8657886 Mfr # F20510560
COMPARE
1500 15 115
$900.24CAD / Each
Velp ARE 5 Analog Stirring Hot Plate, Aluminum; 230V 8657887 Mfr # F20500560
COMPARE
1500 15 230
$900.24CAD / Each