Vaisala Humidity and Temperature Transmitters for Building Automation
Reliable transmitters for basic HVAC humidity measurements
  • User interchangeable INTERCAP® sensor for easy field replacement
  • Easy installation and low maintenance
  • ±3.0% RH accuracy
  • Models available with LCD for local indication
The HMD/W80 series transmitters measure relative humidity and temperature in various building automation applications providing excellent and reliable operation. The interchangeable sensor is easy to replace and requires no recalibration. In addition, the sensor is insensitive to dust, as well as most chemicals. The transmitters require no routine maintenance—a smart choice for HVAC and EMC applications. Available in wall and duct-mount models; both offer simple installation and low maintenance.
$134.64 - $378.85CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Mounting Type Description
Availability Pricing
Vaisala TMW-82 Low-Cost Temperature Transmitter, Wall-Mount RK-37304-01 Mfr # TMW82
COMPARE
55 Wall Low-Cost Temperature Transmitter, Wall-Mount
$134.64CAD / Each
Vaisala HMW-82 Low-Cost Humidity and Temperature Transmitter, Wall-Mount SKU 3730402 RK-37304-02 Mfr # HMW82
COMPARE
55 Wall Low-Cost Humidity and Temperature Transmitter, Wall-Mount
$185.65CAD / Each
Vaisala HMW-88 Low-Cost Humidity and Temperature Transmitter, Wall-Mount and IP-65 Rated RK-37304-03 Mfr # HMW88
COMPARE
55 Wall Low-Cost Humidity and Temperature Transmitter, Wall-Mount and IP-65 Rated
$317.80CAD / Each
Vaisala HMW-88D Humidity/Temperature Transmitter with Display/Wall-Mt/IP-65 RK-37304-04 Mfr # HMW88D
COMPARE
55 Wall Humidity/Temperature Transmitter with Display/Wall-Mt/IP-65
$378.85CAD / Each
Vaisala TMD-82 Low-Cost Temperature Transmitter, Duct-Mount RK-37304-05 Mfr # TMD82
COMPARE
55 Duct Low-Cost Temperature Transmitter, Duct-Mount
$267.70CAD / Each
Vaisala HMD-82 Low-Cost Humidity and Temperature Transmitter, Duct-Mount RK-37304-06 Mfr # HMD82
COMPARE
55 Duct Low-Cost Humidity and Temperature Transmitter, Duct-Mount
$317.80CAD / Each
Vaisala HMD-82D Low-Cost Humidity and Temperature Transmitter with Display, Duct-Mount RK-37304-07 Mfr # HMD82D
COMPARE
55 Duct Low-Cost Humidity and Temperature Transmitter with Display, Duct-Mount
$378.85CAD / Each
Showing 1- 7 of 7