Vacuum Research Pirani and Diaphragm Integral Sensor Gauges
Precision and high resolution of digital electronics plus linear analog outputs for PLCs or data acquisition systems
Pirani sensor gauges 68808-36 to -40 read vacuum from 1 to 1200 mTorr. These compact vacuum instruments offer the precision and high resolution of digital electronics at the same price of analog instruments. The extremely rugged sensor is constructed with a solid, inert noble metal that maintains its calibration over long periods of time. Since no gold is used, these gauges are compatible with both mercury and oil pumps. The low 302°F (125°C) operating temperature of the Pirani-type gauge tube enhances stability, as does the unique temperature compensation network. Diaphragm sensor gauges 68808-42 to -46 read vacuum from 1 to 1500 Torr. This compact gauge features a pure silicon diaphragm that can measure liquid or gas pressure as well as vacuum. Response time is less than 400 milliseconds for full-scale service.
$623.24 - $667.81CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accuracy Resolution Description
Availability Pricing
Vacuum Research 902034 Pirani Sensor, 1 to 1200 mTorr, 1/8" NPT(M) RK-68808-36 Mfr # 902034
COMPARE
±5% of reading or ± 3 mTorr, whichever is larger 1 mTorr Pirani Sensor, 1 to 1200 mTorr, 1/8\" NPT(M)
$667.81CAD / Each
Vacuum Research 902022 Pirani Sensor, 1 to 1200 mTorr, NW-16 RK-68808-38 Mfr # 902022
COMPARE
±5% of reading or ± 3 mTorr, whichever is larger 1 mTorr Pirani Sensor, 1 to 1200 mTorr, NW-16
$623.24CAD / Each