Tygon® F-4040-A Fuel and Lubricant Tubing
Specifically designed to work with hydrocarbons
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
  • Resists swelling and hardening caused by hydrocarbon-based fluids
  • Low extractables help maintain fluid integrity
  • Flexible and easy to install
  • Translucent yellow tubing provides quick identification
  • Ozone and UV resistant
Use the high-performance Tygon® F-4040-A tubing for transferring fuels and industrial lubricants. Specially formulated tubing resists swelling and hardening caused by many hydrocarbon-based fluids that can lead to tubing failure. Low permeation rating makes this tubing perfect for transferring gasoline, kerosene, heating oils, and glycol-based coolants. Highly flexible translucent yellow tubing provides quick identification, easy monitoring of the flow, and fast installation, even in tight places.
$88.26 - $820.68CAD / Pkg of 1
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Tygon F-4040-A Tubing, 3/32" ID x 3/16" OD; 50 Ft 5010591 Mfr # AAG00165
COMPARE
3/32 3/16 3/64
$88.26CAD / Pkg of 1
Tygon F-4040-A Tubing, 1/8" ID x 1/4" OD; 50 Ft 5010592 Mfr # AAG00007
COMPARE
1/8 1/4 1/16
$117.58CAD / Pkg of 1
Tygon F-4040-A Tubing, 3/16" ID x 5/16" OD; 50 Ft 5010593 Mfr # AAG00012
COMPARE
3/16 5/16 1/16
$162.44CAD / Pkg of 1
Tygon F-4040-A Tubing, 1/4" ID x 3/8" OD; 50 Ft 5010594 Mfr # AAG00017
COMPARE
1/4 3/8 1/16
$208.85CAD / Pkg of 1
Tygon F-4040-A Tubing, 5/16" ID x 7/16" OD; 50 Ft 5010595 Mfr # AAG00022
COMPARE
5/16 7/16 1/16
$239.79CAD / Pkg of 1
Tygon F-4040-A Tubing, 3/8" ID x 1/2" OD; 50 Ft 5010596 Mfr # AAG00027
COMPARE
3/8 1/2 1/16
$278.47CAD / Pkg of 1
Tygon F-4040-A Tubing, 1/2" ID x 5/8" OD; 50 Ft 5010597 Mfr # AAG00036
COMPARE
1/2 5/8 1/16
$340.35CAD / Pkg of 1
Tygon F-4040-A Tubing, 3/4" ID x 1" OD; 50 Ft 5010598 Mfr # AAG00053
COMPARE
3/4 1 1/8
$820.68CAD / Pkg of 1
Showing 1- 8 of 8