Tygon® A-60-G Chemical Dispensing Tubing
Specially formulated tubing offers excellent resistance to inorganic fluids
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
  • Outstanding resistance to heat, UV, and ozone
  • Use in temperatures ranging from -60 to 135°C (-75 to 275°F)
  • High flexural fatigue strength—ideal for use in peristaltic pumps
  • Heat sealable—no fittings needed to join tubing
Use the Tygon® A-60-G tubing for industrial cleaning and chemical dispensing applications. It has excellent resistance to inorganic fluids (acids and bases) and will not weaken or crack after exposure to heat, UV, and ozone, outperforming neoprene, EPDM, and other general tubing. Tubing also exhibits high flexural fatigue strength, making it ideal for use in peristaltic pumps.
$78.09 - $696.18CAD / Pkg of 1
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Tygon A-60-G Tubing, 1/16" ID x 3/16" OD; 50 Ft 5010650 Mfr # AFL00003
COMPARE
1/16 3/16 1/16
$78.09CAD / Pkg of 1
Tygon A-60-G Tubing, 1/8" ID x 1/4" OD; 50 Ft 5010651 Mfr # AFL00007
COMPARE
1/8 1/4 1/16
$80.45CAD / Pkg of 1
Tygon A-60-G Tubing, 1/8" ID x 3/8" OD; 50 Ft 5010652 Mfr # AFL00008
COMPARE
1/8 3/8 1/8
$153.23CAD / Pkg of 1
Tygon A-60-G Tubing, 3/16" ID x 5/16" OD; 50 Ft 5010653 Mfr # AFL00012
COMPARE
3/16 5/16 1/16
$86.64CAD / Pkg of 1
Tygon A-60-G Tubing, 1/4" ID x 3/8" OD; 50 Ft 5010654 Mfr # AFL00017
COMPARE
1/4 3/8 1/16
$97.54CAD / Pkg of 1
Tygon A-60-G Tubing, 1/4" ID x 7/16" OD; 50 Ft 5010655 Mfr # AFL00018
COMPARE
1/4 7/16 3/32
$162.44CAD / Pkg of 1
Tygon A-60-G Tubing, 1/4" ID x 1/2" OD; 50 Ft 5010656 Mfr # AFL00019
COMPARE
1/4 1/2 1/8
$232.06CAD / Pkg of 1
Tygon A-60-G Tubing, 5/16" ID x 7/16" OD; 50 Ft 5010657 Mfr # AFL00022
COMPARE
5/16 7/16 1/16
$122.29CAD / Pkg of 1
Tygon A-60-G Tubing, 5/16" ID x 1/2" OD; 50 Ft 5010658 Mfr # AFL00023
COMPARE
5/16 1/2 3/32
$193.38CAD / Pkg of 1
Tygon A-60-G Tubing, 3/8" ID x 1/2" OD; 50 Ft 5010659 Mfr # AFL00027
COMPARE
3/8 1/2 1/16
$137.76CAD / Pkg of 1
Tygon A-60-G Tubing, 3/8" ID x 9/16" OD; 50 Ft 5010660 Mfr # AFL00028
COMPARE
3/8 9/16 3/32
$224.32CAD / Pkg of 1
Tygon A-60-G Tubing, 3/8" ID x 5/8" OD; 50 Ft 5010661 Mfr # AFL00029
COMPARE
3/8 5/8 1/8
$309.41CAD / Pkg of 1
Tygon A-60-G Tubing, 7/16" ID x 9/16" OD; 50 Ft 5010662 Mfr # AFL00032
COMPARE
7/16 9/16 1/16
$255.26CAD / Pkg of 1
Tygon A-60-G Tubing, 1/2" ID x 5/8" OD; 50 Ft 5010663 Mfr # AFL00036
COMPARE
1/2 5/8 1/16
$208.85CAD / Pkg of 1
Tygon A-60-G Tubing, 1/2" ID x 11/16" OD; 50 Ft 5010664 Mfr # AFL00037
COMPARE
1/2 11/16 3/32
$301.68CAD / Pkg of 1
Tygon A-60-G Tubing, 5/8" ID x 13/16" OD; 50 Ft 5010665 Mfr # AFL00045
COMPARE
5/8 13/16 3/32
$433.18CAD / Pkg of 1
Tygon A-60-G Tubing, 5/8" ID x 7/8" OD; 50 Ft 5010666 Mfr # AFL00046
COMPARE
5/8 7/8 1/8
$464.12CAD / Pkg of 1
Tygon A-60-G Tubing, 3/4" ID x 1" OD; 50 Ft 5010667 Mfr # AFL00053
COMPARE
3/4 1 1/8
$534.10CAD / Pkg of 1
Tygon A-60-G Tubing, 1" ID x 1-1/4" OD; 50 Ft 5010668 Mfr # AFL00062
COMPARE
1 1 1/4 1/8
$696.18CAD / Pkg of 1
Showing 1- 19 of 19