Troemner Class-F Metric Stainless Steel Cylindrical Test Weights
Stainless Steel Test Weights are primarily used to test commercial weighing devices by state and local weights and measures officials, device installers, and service technicians.
Stainless Steel Cylindrical Weights are legal for trade, which means their design meets specifications as outlined in NIST Handbook 105-1 and are approved to be used on legal for trade devices within the United States. Weights 5 kg/10 lb and below must be made of stainless steel of quality no lower than 300 series. Adjusting cavities are typically on the top of the weight and must be tamper resistant.
Stainless Steel Class F Test Weights may be used to test most accuracy Class III scales, all scales of Class IIIL or IIII, and scales not marked with a class designation. Class F is also acceptable
for use in verifying scales that have 0.1% accuracy or lower. Class F Test Weights are primarily used to test commercial weighing devices by state and local weights and measures officials, device installers, and service technicians. Suggested markets for Stainless Steel Test Weights include, but are not limited to, Hospitals and Food Services.
$59.08 - $551.78CAD / Each
31 variations of this product are available.
See All Product Options Below
31 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Certification Mass Size (g) Mass Size (mg)
Availability Pricing
Troemner Test Weight 1g Class F with Traceable Certificate, Cyl Indrical with Groove 0102346 Mfr # 1334T
COMPARE
NIST 1 1000
$170.91CAD / Each
Troemner Test Weight 2g Class F with Traceable Certificate, Cyl Indrical with Groove 0102347 Mfr # 1332T
COMPARE
NIST 2 2000
$170.91CAD / Each
Troemner Test Weight 3 G Class F with Traceable Certificate, Cyl Indrical with Groove 0102348 Mfr # 1330T
COMPARE
NIST 3 3000
$170.91CAD / Each
Troemner Test Weight 10g Class F with Traceable Certificate, Cyl Indrical with Groove 0102349 Mfr # 1326T
COMPARE
NIST 10 10000
$184.91CAD / Each
Troemner Test Weight 20g Class F with Traceable Certificate, Cyl Indrical with Groove 0102350 Mfr # 1324T
COMPARE
NIST 20 20000
$184.91CAD / Each
Troemner Test Weight 30g Class F with Traceable Certificate, Cyl Indrical with Groove 0102351 Mfr # 1322T
COMPARE
NIST 30 30000
$184.91CAD / Each
Troemner Test Weight 50g Class F with Traceable Certificate, Cyl Indrical with Groove 0102352 Mfr # 1320T
COMPARE
NIST 50 50000
$187.86CAD / Each
Troemner Test Weight 100g Class F with Traceable Certificate 0102354 Mfr # 1318T
COMPARE
NIST 100 100000
$194.49CAD / Each
Troemner Test Weight 200g Class F with Traceable Certificate 0102356 Mfr # 1316T
COMPARE
NIST 200 200000
$215.85CAD / Each
Troemner Test Weight 300g Class F with Traceable Certificate 0102357 Mfr # 1314T
COMPARE
NIST 300 300000
$246.79CAD / Each
Troemner Test Weight 400g Class F with Traceable Certificate 0102358 Mfr # 1313T
COMPARE
NIST 400 400000
$295.05CAD / Each
Troemner Test Weight 500g Class F with Traceable Certificate 0102360 Mfr # 1312T
COMPARE
NIST 500 500000
$298.36CAD / Each
Troemner Test Weight 1000g Class F with Traceable Certificate 0102362 Mfr # 1310T
COMPARE
NIST 1000 1000000
$321.94CAD / Each
Troemner Test Weight 2000g Class F with Traceable Certificate 0102364 Mfr # 1308T
COMPARE
NIST 2000 2000000
$409.97CAD / Each
Troemner Test Weight 3000g Class F , Traceable Certificate, Cyl Indrical, Groove 0102366 Mfr # 1306T
COMPARE
NIST 3000 3000000
$503.53CAD / Each
Troemner Test Weight 5000g Class F with Traceable Certificate 0102368 Mfr # 1302T
COMPARE
NIST 5000 5000000
$551.78CAD / Each
Troemner 1328T 5 g Class F Test Weight with Traceable Certificate Cylindrical with Groove 0103376 Mfr # 1328T
COMPARE
NIST 5 5000
$170.91CAD / Each
Troemner 1302 5000 g Class F Test Weight with No Certificate Cylindrical with Groove 0103377 Mfr # 1302
COMPARE
None 5000 5000000
$455.28CAD / Each
Troemner 1306 3000 g Class F Test Weight with No Certificate Cylindrical with Groove 0103378 Mfr # 1306
COMPARE
None 3000 3000000
$382.34CAD / Each
Troemner 1308 2000 g Class F Test Weight with No Certificate Cylindrical with Groove 0103379 Mfr # 1308
COMPARE
None 2000 2000000
$270.37CAD / Each
Showing 1 - 20 of 31