Trajan Taper FocusLiner Inlet Liners

Spot the Difference in the SGE Lineup!
  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
Bottom taper minimizes contact with metal parts of the inlet and focuses sample onto the column head. Top taper aids in minimizing sample flashback
$88.99 - $1,337.84CAD / Pkg of 5

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 092003 Split/Splitless Tapered Focus Liner, Compatible with Agilent Systems, 78.5 mm L; 5/Pk 1600924 Mfr # 092003
COMPARE
$345.88CAD / Pkg of 5
Trajan SGE 09200301 Split/Splitless Tapered Focus Liner, Compatible with Agilent Systems, 78.5 mm L; 1/Pk 1600925 Mfr # 09200301
COMPARE
$88.99CAD / Pkg of 1
Trajan SGE 092011 Split/Splitless Tapered Focus Liner, Compatible with Agilent Systems, 78.5 mm L; 25/Pk 1600935 Mfr # 092011
COMPARE
$1,337.84CAD / Pkg of 25
Trajan SGE 092058 Tapered Focus Liners, Caompatible with Shimadzu 2010/17A, 3.4 mm ID , 95 mm L; 5/Pk 1600985 Mfr # 092058
COMPARE
$372.40CAD / Pkg of 5
Trajan SGE 092111 Split/Splitless Tapered Fast Focus Liner, Compatible with Agilent Systems, 78.5 mm L; 5/Pk 1601046 Mfr # 092111
COMPARE
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat