Trajan SGE 09200301 Split/Splitless Tapered Focus Liner, Compatible with Agilent Systems, 78.5 mm L; 1/Pk

Representative image only.

Trajan SGE 09200301 Split/Splitless Tapered Focus Liner, Compatible with Agilent Systems, 78.5 mm L; 1/Pk

Trajan | Mfr: 09200301| Item: 1600925

Spot the Difference in the SGE Lineup!

$88.99CAD / Pkg of 1

$88.99CAD / Pkg of 1
Product Description

Key Features

  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
  • 6 month warranty

More About this Item

Bottom taper minimizes contact with metal parts of the inlet and focuses sample onto the column head. Top taper aids in minimizing sample flashback

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

6 Month Warranty
Specifications
  • DescriptionTrajan SGE Split/Splitless Tapered Focus Liner, Compatible with Agilent Systems, 78.5 mm L; 1/Pk
  • Warranty6 month warranty
Reviews
Q&A