Trajan Straight Chromatography Liners

Spot the Difference in the SGE Lineup!
  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
Straight liners facilitate higher split flows
$207.38 - $748.48CAD / Pkg of 5

12 variations of this product are available.

See All Product Options Below
12 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 092004 Splitless straight-through Liner, Compatible with Agilent Systems, 78.5 mm L; 5/Pk 1600926 Mfr # 092004
COMPARE
$223.59CAD / Pkg of 5
Trajan SGE 092016 Direct Straight-through Liner, Compatible with Agilent Systems, 1.2 mm ID, 78.5 mm L; 5/Pk 1600938 Mfr # 092016
COMPARE
$207.38CAD / Pkg of 5
Trajan SGE 092027 Inlet Liner, Compatible with Varian 1093/1094(Restriction = 0.25mm), 0.5 mm ID; 5/Pk 1600954 Mfr # 092027
COMPARE
$371.29CAD / Pkg of 5
Trajan SGE 092034 Inlet Liners, Varian 1093/1094 (Restriction=0.50mm), 0.8 mm ID, 54 mm L; 5/Pk 1600962 Mfr # 092034
COMPARE
$365.77CAD / Pkg of 5
Trajan SGE 092089 SPME Liner, Compatible with Shimadzu 17A, 0.75 mm ID, 95 mm L; 5/Pk 1601023 Mfr # 092089
COMPARE
$212.54CAD / Pkg of 5
Trajan SGE 092100 Split/Splitless Single Tapered Liner, Compatible with PerkinElmer Systems, 4 mm ID; 5/Pk 1601039 Mfr # 092100
COMPARE
$248.27CAD / Pkg of 5
Trajan SGE 092142 PTV Trace 2000 Liner, Compatible with Thermo Trace Liner, 2 mm ID, 120 mm L; 5/Pk 1601054 Mfr # 092142
COMPARE
$386.40CAD / Pkg of 5
Trajan SGE 092150 Split Straight-through Liner, Compatible with Thermo Trace Liner, 5 mm ID; 5/Pk 1601062 Mfr # 092150
COMPARE
$252.69CAD / Pkg of 5
Trajan SGE 092220 Split Liners with Quartz Wool, Compatible with Agilent Systems, 4 mm ID, 78.5 mm L; 25/Pk 1601072 Mfr # 092220
COMPARE
$748.48CAD / Pkg of 25
Trajan SGE 092222 Split straight-through Liner, Compatible with Agilent systems, 4 mm ID, 78.5 mm L; 25/Pk 1601074 Mfr # 092222
COMPARE
$616.61CAD / Pkg of 25
Trajan SGE 092236 Packed Column Liner, Compatible with PerkinElmer Systems, 112 mm L; 5/Pk 1601083 Mfr # 092236
COMPARE
$509.06CAD / Pkg of 5
Trajan SGE 092500 Dani Straight-through Liner with quartz wool, 4 mm ID, 70 mm L; 5/Pk 1601125 Mfr # 092500
COMPARE
$378.66CAD / Pkg of 5
Showing 1- 12 of 12

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat