Trajan SGE 092027 Inlet Liner, Compatible with Varian 1093/1094(Restriction = 0.25mm), 0.5 mm ID; 5/Pk

Representative image only.

Trajan SGE 092027 Inlet Liner, Compatible with Varian 1093/1094(Restriction = 0.25mm), 0.5 mm ID; 5/Pk

Trajan | Mfr: 092027| Item: 1600954

Spot the Difference in the SGE Lineup!

$371.29CAD / Pkg of 5

$371.29CAD / Pkg of 5
Product Description

Key Features

  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
  • 6 month warranty

More About this Item

Straight liners facilitate higher split flows

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

6 Month Warranty
Specifications
  • DescriptionTrajan SGE Inlet Liner, Compatible with Varian 1093/1094(Restriction = 0.25mm), 0.5 mm ID; 5/Pk
  • Warranty6 month warranty
Reviews
Q&A