Trajan SGE Taper/Gooseneck Inlet Liners

Spot the Difference in the SGE Lineup!
  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
Addition of f.e.l.t. increases lifetime, impreoves repeatability, sensitivity, reproduceibility, and ability to handle dirty matrices
$58.20 - $957.70CAD / Pkg of 5

14 variations of this product are available.

See All Product Options Below
14 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 092013 Splitless Liners with Recessed Gooseneck, Compatible with Agilent Systems, 78.5 mm L; 5/Pk 1600936 Mfr # 092013
COMPARE
$254.16CAD / Pkg of 5
Trajan SGE 09201701 Split/Splitless Liner with single taper, Compatible with Agilent Systems, 78.5 mm L; 1/Pk 1600941 Mfr # 09201701
COMPARE
$58.20CAD / Pkg of 1
Trajan SGE 092019 Liner with Tappered quartz wool, Compatible with Agilent Systems, 78.5 mm L; 5/Pk 1600944 Mfr # 092019
COMPARE
$238.32CAD / Pkg of 5
Trajan SGE 09201901 Liner with Tappered quartz wool, Compatible with Agilent Systems, 78.5 mm L; 1/Pk 1600945 Mfr # 09201901
COMPARE
$61.88CAD / Pkg of 1
Trajan SGE 092021 Split Liners with Quartz Wool, Compatible with Varian 1075/1077, 4 mm ID, 72 mm L; 5/Pk 1600946 Mfr # 092021
COMPARE
$367.61CAD / Pkg of 5
Trajan SGE 092061 Inlet Liners, Gooseneck with Quartz Wool, Compatible with Shimadzu 2010/17A,3.4mm ID; 5/Pk 1600991 Mfr # 092061
COMPARE
$249.00CAD / Pkg of 5
Trajan SGE 092071 Single Tapered Liner, Compatible with Shimadzu 2010/17A, 3.4 mm ID, 95 mm L; 5/Pk 1601005 Mfr # 092071
COMPARE
$373.50CAD / Pkg of 5
Trajan SGE 092077 Liner with Upper Mid-Gooseneck, Compatible with Shimadzu 2010/17A, 3.4 mm ID; 5/Pk 1601007 Mfr # 092077
COMPARE
$295.05CAD / Pkg of 5
Trajan SGE 092085 Liner with Mid-Gooseneck, Compatible with Shimadzu 17A, 3.4 mm ID, 95 mm L; 5/Pk 1601015 Mfr # 092085
COMPARE
$253.42CAD / Pkg of 5
Trajan SGE 092087 Single Tapered Liner, Compatible with Shimadzu 17A, 2.6 mm ID, 95 mm L; 5/Pk 1601020 Mfr # 092087
COMPARE
$321.57CAD / Pkg of 5
Trajan SGE 092091 Split/Splitless Liner with Gooseneck, Compatible with PerkinElmer Systems, 3 mm ID; 5/Pk 1601025 Mfr # 092091
COMPARE
$241.27CAD / Pkg of 5
Trajan SGE 092218 Liner with Tapered Quartz Wool, Compatible with Agilent Systems, 4 mm ID, 78.5 mm L; 25/Pk 1601070 Mfr # 092218
COMPARE
$947.39CAD / Pkg of 25
Trajan SGE 092223 Recessed Gooseneck with quartz wool, Compatible with Agilent Systems, 78.5 mm L; 25/Pk 1601075 Mfr # 092223
COMPARE
$957.70CAD / Pkg of 25
Trajan SGE 092229 Split/Splitless Tapered Liner, Compatible with Agilent Systems, 4 mm ID, 78.5 mm L; 25/Pk 1601078 Mfr # 092229
COMPARE
$934.13CAD / Pkg of 25
Showing 1- 14 of 14

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat