Trajan SGE 09201901 Liner with Tappered quartz wool, Compatible with Agilent Systems, 78.5 mm L; 1/Pk

Representative image only.

Trajan SGE 09201901 Liner with Tappered quartz wool, Compatible with Agilent Systems, 78.5 mm L; 1/Pk

Trajan | Mfr: 09201901| Item: 1600945

Spot the Difference in the SGE Lineup!

$61.88CAD / Pkg of 1

$61.88CAD / Pkg of 1
Product Description

Key Features

  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
  • 6 month warranty

More About this Item

Addition of f.e.l.t. increases lifetime, impreoves repeatability, sensitivity, reproduceibility, and ability to handle dirty matrices

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

6 Month Warranty
Specifications
  • DescriptionTrajan SGE Liner with Tappered quartz wool, Compatible with Agilent Systems, 78.5 mm L; 1/Pk
  • Warranty6 month warranty
Reviews
Q&A