Trajan Graphite GC Ferrules
Recommended applications are column to injector and column to FID connections. Unsuitable for connecting columns to mass spectrometers
  • Easy to use
  • Forms a reliable seal
  • Reusable; soft material; forms a soft grip to the capillary column
Variety of hole sizes available for 1/16", 1/8" and 1/4" fittings. 450°C temperature limit.
$64.39 - $170.91CAD / Pkg of 10
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Trajan Replacement Graphite Short Ferrules, Compatible with 1/4" OD Columns; 10/Pk 1600715 Mfr # 072601
COMPARE
Trajan SGE Replacement Graphite Ferrules, Compatible with 1/4" OD Columns; 10/Pk
$64.39CAD / Pkg of 10
Trajan Replacement Graphite Short Ferrules, Compatible with 1/8" OD Columns; 10/Pk 1600716 Mfr # 072602
COMPARE
Trajan SGE Replacement Graphite Ferrules, Compatible with 1/8" OD Columns; 10/Pk
$64.39CAD / Pkg of 10
Trajan Replacement Graphite Short Ferrules, Compatible with 1/16" OD Columns; 10/Pk 1600717 Mfr # 072603
COMPARE
Trajan SGE Replacement Graphite Ferrules, Compatible with 1/16" OD Columns; 10/Pk
$65.57CAD / Pkg of 10
Trajan Graphite Short Reducing Ferrule With 6 Mm Hole For 1/4" Fittings, Pk10 1600718 Mfr # 072604
COMPARE
Trajan SGE Graphite Short Reducing Ferrule with 6 mm Hole for 1/4" Fittings; 10/Pk
$71.31CAD / Pkg of 10
Trajan Graphite Short Reducing Ferrule With 1.2 Mm Hole For 1/16" Fittings, Pk10 1600720 Mfr # 072606
COMPARE
Trajan SGE Graphite Reducing Short Ferrule for 1/16" Fittings, 1.2 mm Hole; 10/Pk
$72.05CAD / Pkg of 10
Trajan Graphite Short Reducing Ferrule With 1 Mm Hole For 1/16" Fittings, Pk10 1600721 Mfr # 072607
COMPARE
Trajan Graphite short reducing ferrule with 0.8 mm hole for 1/16" fittings, PK10
$71.46CAD / Pkg of 10
Trajan Graphite Short Reducing Ferrule With 0.8 Mm Hole For 1/16" Fittings, Pk10 1600722 Mfr # 072608
COMPARE
Trajan SGE Graphite Reducing Short Ferrule for 1/16" Fittings, 0.8 mm Hole; 10/Pk
$72.05CAD / Pkg of 10
Trajan Graphite Ferrule With 0.8 Mm Hole For Gc14A, Gc17A, Gc2010 And Gc2014 Detector/Injectors 1600724 Mfr # 0726082
COMPARE
Trajan Graphite ferrule with 0.8 mm hole for GC14A, GC17A, GC2010 and GC2014 detector/injectors
$170.91CAD / Pkg of 10
Trajan Graphite Short Ferrule With 1/2" Hole For 1/2" Fittings, Pk10 1600726 Mfr # 072613
COMPARE
Trajan Graphite short ferrule with 1/2" hole for 1/2" fittings, PK10
$84.72CAD / Pkg of 10
Trajan Graphite Ferrule, Compatible with Carlo Erba; 10/Pk 1600727 Mfr # 072614
COMPARE
Trajan Graphite ferrule with 0.8 mm hole for Carlo Erba detector/injectors
$65.57CAD / Pkg of 10
Trajan Graphite Short Ferrule With 3/8" Hole For 3/8" Fittings, Pk10 1600728 Mfr # 072615
COMPARE
Trajan Graphite short ferrule with 3/8" hole for 3/8" fittings, PK10
$84.57CAD / Pkg of 10
Trajan Graphite Reducing Ferrule With 6 Mm Hole For 1/4" Fittings, Pk10 1600733 Mfr # 0726212
COMPARE
Trajan Graphite reducing ferrule with 6 mm hole for 1/4" fittings, PK10
$65.57CAD / Pkg of 10
Trajan Graphite Ferrule With 1/8" Hole For 1/8" Od Packed Columns, Pk10 1600735 Mfr # 072622
COMPARE
Trajan Graphite ferrule with 1/8" hole for 1/8" OD packed columns, PK10
$65.57CAD / Pkg of 10
Trajan Graphite Straight Ferrule with 1/16" Hole for 1/16" Fittings,  10/Pk 1600736 Mfr # 072623
COMPARE
Trajan Graphite ferrule with 1/16" hole for 1/16" fittings, PK10
$65.57CAD / Pkg of 10
Trajan Graphite Ferrules, Compatible with PerkinElmer 1/8" Nut ; 10/Pk 1600737 Mfr # 072624
COMPARE
Trajan Graphite ferrules with 0.5 mm hole for use with 1/8" nuts on PerkinElmer injectors and detectors, PK10
$65.57CAD / Pkg of 10
Trajan Graphite Reducing Ferrule for 1/16" Fittings, 1 mm Hole,  10/Pk 1600738 Mfr # 072625
COMPARE
Trajan Graphite ferrule with 1.0 mm hole for 1/16" fittings, PK10
$65.57CAD / Pkg of 10
Trajan Graphite Ferrules, Compatible with PerkinElmer 1/8" Nut and 1.0 mm hole; 10/Pk 1600741 Mfr # 0726280
COMPARE
Trajan Graphite ferrules with 1.0 mm hole for use with 1/8" nuts on PerkinElmer injectors and detectors, PK10
$65.57CAD / Pkg of 10
Trajan Graphite Ferrule for 1/8" Fittings, 1/16" Hole,10/Pk 1600744 Mfr # 0726283
COMPARE
Trajan Graphite ferrule with 1/16" hole for 1/8" fittings, PK10
$65.57CAD / Pkg of 10
Trajan Graphite Short Reducing Ferrule With 0.5 Mm Hole For Detector/Injectors (Not For Msd Interface Connections), Pk10 1600746 Mfr # 072635
COMPARE
Trajan Graphite short reducing ferrule with 0.5 mm hole for detector/injectors (not for MSD interface connections), PK10
$65.27CAD / Pkg of 10
Trajan Graphite Short Reducing Ferrule With 0.8 Mm Hole For Detector/Injectors (Not For Msd Interface Connections), Pk10 1600747 Mfr # 072636
COMPARE
Trajan Graphite short reducing ferrule with 0.8 mm hole for detector/injectors (not for MSD interface connections), PK10
$65.27CAD / Pkg of 10
Showing 1 - 20 of 24